Skip to Content
Merck
All Photos(1)

Key Documents

T7830

Sigma-Aldrich

TFLLR-NH2 trifluoroacetate salt

>98% (HPLC)

Synonym(s):

L-Threonyl-L-phenylalanyl-L-leucyl-L-leucyl- L-argininamide trifluoroacetate salt, Thr-Phe-Leu-Leu-Arg-NH2 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$185.00
25 MG
$729.00

$185.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$185.00
25 MG
$729.00

About This Item

Empirical Formula (Hill Notation):
C31H53N9O6 · xC2HF3O2
CAS Number:
Molecular Weight:
647.81 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$185.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>98% (HPLC)

form

lyophilized powder

color

white to tan

solubility

H2O: >2 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)[C@@H](C)O)C(=O)N[C@@H](CCCNC(N)=N)C(N)=O

InChI

1S/C31H53N9O6.C2HF3O2/c1-17(2)14-22(27(43)37-21(26(33)42)12-9-13-36-31(34)35)38-28(44)23(15-18(3)4)39-29(45)24(16-20-10-7-6-8-11-20)40-30(46)25(32)19(5)41;3-2(4,5)1(6)7/h6-8,10-11,17-19,21-25,41H,9,12-16,32H2,1-5H3,(H2,33,42)(H,37,43)(H,38,44)(H,39,45)(H,40,46)(H4,34,35,36);(H,6,7)/t19-,21+,22+,23+,24+,25+;/m1./s1

InChI key

QVNWOGSDGQDGHP-MKVNCOEFSA-N

Amino Acid Sequence

Thr-Phe-Leu-Leu-Arg-NH2

Application

TFLLR-NH2 trifluoroacetate salt has been used as a protease-activated receptor-1 (PAR-1) agonist to study its effects on hippocampal CA1 pyramidal neurons and Swell1 knock out mice (cKO).[1]

Biochem/physiol Actions

TFLLR-NH2 is a protease-activated receptor (PARs) agonist which is more selective to PAR-1 than SFLLRN-NH2.
TFLLR-NH2 is a protease-activated receptor (PARs) agonist which is more selective to PAR-1 than SFLLRN-NH2. Protease-activated receptors (PARs) are present on various organs including, plateles, mast cell, gallblader, oesophagus etc, and regulate various physiological processes including human platelet aggregation, vascular contraction/relaxation, and an increase in endothelial permeability. Recent papers indicated that PAR′s are also involved in sensory processing. Specificly PAR ligands enhance glutamatergic excitatory transmission in substantia gelatinosa (SG) neurons of adult rat spinal cord slices.

Preparation Note

TFLLR-NH2 trifluoroacetate is soluble in water at a concentration that is greater than 2 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Glutamate-releasing SWELL1 channel in astrocytes modulates synaptic transmission and promotes brain damage in stroke
Yang J, et al.
Neuron, 102(4), 813-827 (2019)
Hui Sun et al.
American journal of physiology. Lung cellular and molecular physiology, 318(1), L192-L199 (2019-10-31)
We evaluated the mechanisms underlying protease-activated receptor 1 (PAR1)-mediated activation of nodose C-fibers in mouse lungs. The PAR1-induced action potential discharge at the terminals was strongly inhibited in phospholipase C-β3 (PLCβ3)-deficient animals. At the level of the cell soma, PAR1
Junhua Yang et al.
Neuron, 102(4), 813-827 (2019-04-16)
By releasing glutamate, astrocytes actively regulate synaptic transmission and contribute to excitotoxicity in neurological diseases. However, the mechanisms of astrocytic glutamate release have been debated. Here, we report non-vesicular release of glutamate through the glutamate-permeable volume-regulated anion channel (VRAC). Both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service