Skip to Content
Merck

Skip To

T7799

Tumor Necrosis Factor-β human

>95% (SDS-PAGE), recombinant, expressed in E. coli, powder, suitable for cell culture | mammalian: suitable

Synonym(s):

TNF-β

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Biological source:
human
Recombinant:
expressed in E. coli
Assay:
>95% (SDS-PAGE)
Form:
powder
Mol wt:
protein 18.8 kDa
Impurities:
≤1 EU/μg
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Tumor Necrosis Factor-β human, TNF-β, recombinant, expressed in E. coli, powder, suitable for cell culture

biological source

human

Quality Segment

recombinant

expressed in E. coli

assay

>95% (SDS-PAGE)

form

powder

potency

4-20 pg/mL Activity

quality

endotoxin tested

mol wt

protein 18.8 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

Biochem/physiol Actions

Tumor necrosis factor-β (TNF-β), also known as lymphotoxin-α (LT-α) and cytotoxic factor, is a 25 kDa glycoprotein that is tightly regulated by lymphocytes. TNF-β and TNF-α share a 28% amino acid sequence identity, and as soluble forms, both can exist in solution as homotrimers, activate the same TNF receptors (TNFR1 and TNFR2) and elicit many of the same bioactivities. Unlike TNF-α, TNF-β can also form heterotrimers with one or two molecules of the related membrane-bound lymphotoxin-β (LT-β). Reportedly, LTα1/β2, the predominant heterotrimer, binds and activates only the LT-β receptor, LT-βR. The other heterotrimer, LT-α2/β1 binds and activates TNFR1 and TNFR2 but not LT-βR. TNF-β is expressed in activated T, B and NK cells and in certain transformed cells. Genes for TNF-β, LT-β and TNF-α are tightly linked near each other within the MHC. TNF-β appears to play many of the same biological roles as TNF-α, but the membrane-bound form with LT-β is critical for normal lymphoid organ development. Genetic polymorphisms in TNF-α and TNF-β have been linked to certain pathological conditions, including myasthenia gravis. At the amino acid level, human and mouse TNF-β are 74% homologous and human and mouse LT-β are 80% homologous.
Tumor necrosis factor-β, also known as lymphotoxin-α (LT-α) and cytotoxic factor, is a 25 kDa glycoprotein that is tightly regulated by lymphocytes.

Physical form

lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein

Analysis Note

The biological activity is measured in a cytotoxicity assay using the TNF-suspectible murine L929 cell line in the presence of actinomycin D.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions






Global Trade Item Number

SKUGTIN
T7799-10UG04061837378645

Questions

Reviews

No rating value

Active Filters