Skip to Content
Merck
All Photos(2)

Key Documents

T7567

Sigma-Aldrich

Tributylphosphine solution

200 mM (in N-methyl-2-pyrrolidinone), liquid

Synonym(s):

TBP

Sign Into View Organizational & Contract Pricing

Select a Size

10 VIALS
$140.00

$140.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
10 VIALS
$140.00

About This Item

Empirical Formula (Hill Notation):
C12H27P
CAS Number:
Molecular Weight:
202.32
MDL number:
UNSPSC Code:
41105323
PubChem Substance ID:
NACRES:
NA.56

$140.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

form

liquid

Quality Level

concentration

200 mM (in N-methyl-2-pyrrolidinone)

SMILES string

CCCCP(CCCC)CCCC

InChI

1S/C12H27P/c1-4-7-10-13(11-8-5-2)12-9-6-3/h4-12H2,1-3H3

InChI key

TUQOTMZNTHZOKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

TBP is an uncharged reducing agent for disulfide bonds of cysteine residues. This reagent is ideally suited for preparation of protein samples prior to IEF and 2D electrophoresis. Reduction with TBP and subsequent alkylation with iodoacetamide improves resolution, allows greater sample loads, and therefore, improves visualization of low abundance proteins

Packaging

Sealed ampule of 0.5 mL

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stuart Thomson et al.
Clinical & experimental metastasis, 25(8), 843-854 (2008-08-13)
NSCLC cells with a mesenchymal phenotype have shown a marked reduction in sensitivity to EGFR inhibitors, though the molecular rationale has remained obscure. Here we find that in mesenchymal-like tumor cells both tyrosine phosphorylation of EGFR, ErbB2, and ErbB3 signaling
T Oe et al.
Journal of chromatography. B, Biomedical sciences and applications, 708(1-2), 285-289 (1998-07-08)
A method was developed for the simultaneous determination of gamma-glutamylglutathione (gamma-GluGSH) and other low-molecular-mass thiol compounds (cysteine, cysteamine, homocysteine, cysteinylglycine, gamma-glutamylcysteine, glutathione and N-acetylcysteine) using high-performance liquid chromatography combined with precolumn fluorescence labeling with ammonium 7-fluorobenzo-2-oxa-1,3-diazole-4-sulphonate (SBD-F). These SBD-labeled thiol
Tien Q Pham et al.
The Journal of organic chemistry, 70(16), 6369-6377 (2005-07-30)
The phosphine-catalyzed [3 + 2]-cycloaddition of 5-methylenehydantoins 4 with the ylides 5, derived from addition of tributylphosphine to the 2-butynoic acid derivatives, 6a-d, gives spiro-heterocyclic products. The camphor sultam derivative 6b gives optically active products. Noteable was that the ylides
Toon Verheyen et al.
Nucleic acids research, 43(12), 5771-5784 (2015-05-21)
Protein phosphatase-1 (PP1) is a key regulator of transcription and is targeted to promoter regions via associated proteins. However, the chromatin binding sites of PP1 have never been studied in a systematic and genome-wide manner. Methylation-based DamID profiling in HeLa
Claudia Cattoglio et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(18), E2317-E2326 (2015-04-23)
The embryonic stem cell (ESC) state is transcriptionally controlled by OCT4, SOX2, and NANOG with cofactors, chromatin regulators, noncoding RNAs, and other effectors of signaling pathways. Uncovering components of these regulatory circuits and their interplay provides the knowledge base to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service