Skip to Content
Merck
All Photos(2)

Key Documents

T7527

Sigma-Aldrich

Tris-Borate-EDTA buffer

BioReagent, for molecular biology, 5x concentrate, DNase and RNase, none detected, powder blend, suitable for electrophoresis

Synonym(s):

TBE buffer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

Molecular Biology
for molecular biology

product line

BioReagent

form

powder blend

impurities

DNase and RNase, none detected

pH

8.1-8.5 (5 ×)

solubility

water: 85.1 g/L, clear, colorless

suitability

suitable for electrophoresis

application(s)

diagnostic assay manufacturing

SMILES string

OB(O)O.NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.C4H11NO3.BH3O3/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8;2-1(3)4/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2;2-4H

InChI key

OSBLTNPMIGYQGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ready for use in gel electrophoresis after dilution to working concentrations.
TBE running buffer is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel. TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids.
Dilution of the TBE stock concentrates to a 1× TBE running buffer results in a buffer containing 89 mM Tris-borate and 2 mM EDTA, pH 8.3. The 5× or 10× stocks may also be added to an acrylamide/bis-acrylamide stock solution for making the PAGE gel. Applied voltages of less than 5 V/cm (distance between the electrodes of the unit) are recommended for maximum resolution.

Packaging

1L poly bottle contains a powder blend that can be dissolved and reconstituted within the bottle to prepare one liter of a 5× concentrate.
4L poly bottle contains a powder blend that can be dissolved and reconstituted to prepare four liters of a 5× concentrate.


Preparation Note

Prepared with Biotechnology Performance Certified Trizma base (Product Code T6066) and Molecular Biology Reagents boric acid (Product Code B6768) and EDTA disodium salt (Product Code E5134).

Reconstitution

Produces a 5× concentrate (0.445 M Tris-borate, 10 mM EDTA, pH 8.3) after dissolving with the indicated amount of water.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miyuki Watabe et al.
The Ulster medical journal, 77(3), 168-174 (2008-10-30)
In Northern Ireland over the last 7 years, there is a mean of 41.9 laboratory reports per annum of human gastrointestinal infection (range 19-54) caused by Escherichia coli O157:H7. In the preceding years 1992-1996, reports were 5.4 per annum, whereas
Josep Balart et al.
Radiation oncology (London, England), 6, 6-6 (2011-01-18)
Radiation-induced DNA double-strand break (DSB) repair can be tested by using pulsed-field gel electrophoresis (PFGE) in agarose-encapsulated cells. However, previous studies have reported that this assay is impaired by the spontaneous DNA breakage in this medium. We investigated the mechanisms
Seung-min Park et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(37), 15549-15554 (2009-09-01)
Nanofluidics represents a promising solution to problems in fields ranging from biomolecular analysis to optical property tuning. Recently a number of simple nanofluidic fabrication techniques have been introduced that exploit the deformability of elastomeric materials like polydimethylsiloxane (PDMS). These techniques

Protocols

TAE and TBE are both used as running buffers for nucleic acid electrophoresis but have some important differences. Review our recipes and video to give your application the best chance of success.

Related Content

Nancy-520 for DNA Detection and Quantitation

Questions

  1. I have a bottle of your Tris borate- EDTA buffer substance (powder) it is 5 x concentrate, I need to know how much powder to add to 1000ml water please?

    1 answer
    1. This product is pre-aliquoted with the appropriate amount to generate 1 or 4 L of a 5X concentrate. The 1L poly bottle contains a powder blend that can be dissolved and reconstituted within the bottle to prepare one liter of a 5× concentrate. The 4L poly bottle contains a powder blend that can be dissolved and reconstituted to prepare 4L of a 5× concentrate.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service