Skip to Content
Merck
All Photos(2)

Documents

T7375

Sigma-Aldrich

Thionicotinamide adenine dinucleotide

≥90%

Synonym(s):

Thionicotinamide-DPN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H27N7O13SP2
CAS Number:
Molecular Weight:
679.49
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥90%

storage temp.

−20°C

SMILES string

NC(=S)C1=CC=C[N](=C1)C2OC(COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3O)n4cnc5c(N)ncnc45)C(O)C2O

InChI

1S/C21H28N7O13P2S/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(40-21)6-38-43(35,36)41-42(33,34)37-5-10-13(29)15(31)20(39-10)27-3-1-2-9(4-27)18(23)44/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H2,23,44)(H,33,34)(H,35,36)(H2,22,24,25)

InChI key

JFOSDPGFZXVRDA-UHFFFAOYSA-N

Application

Thionicotinamide adenine dinucleotide has been used as a substrate analog in S-adenosyl-l-homocysteine (AdoHcy) hydrolases (SAHH) inhibition assay and in NAD+ glycohydrolase activity.

Biochem/physiol Actions

Thionicotinamide adenine dinucleotide (sNAD) is an analog of coenzyme nicotinamide adenine dinucleotide (NAD) and NADH. sNAD is a potential inhibitor of NAD+ kinase. NAD+ kinase modulates NAD levels, contributing to cytotoxicity in cancer cells.

Linkage

Analog of NAD

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Circular dichroic properties and conformation of thionicotinamide dinucleotides
JOPPICH-KUHN R and LUISI PL
European Journal of Biochemistry, 83(2), 587-592 (1978)
ALT reagent with thionicotinamide adenine dinucleotide.
P A Dolan et al.
Clinical chemistry, 35(9), 1857-1858 (1989-09-01)
William E Karsten et al.
Biochemistry, 41(40), 12193-12199 (2002-10-03)
Tartrate dehydrogenase catalyzes the divalent metal ion- and NAD-dependent oxidative decarboxylation of D-malate to yield CO(2), pyruvate, and NADH. The enzyme also catalyzes the metal ion-dependent oxidation of (+)-tartrate to yield oxaloglycolate and NADH. pH-rate profiles and isotope effects were
[Simultaneous assay of serum lactate dehydrogenase activities depending on NAD (N) and thio-NAD (T), and the clinical significance of N/T values].
M Kawai
Nihon Shokakibyo Gakkai zasshi = The Japanese journal of gastro-enterology, 83(11), 2458-2458 (1986-11-01)
NAD+ kinase as a therapeutic target in cancer
Tedeschi PM, et al.
Clinical Cancer Research, 22(21), 5189-5195 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service