Skip to Content
Merck
All Photos(1)

Key Documents

T6568

Sigma-Aldrich

Tyrphostin RG 13022

≥98%

Synonym(s):

2-(3,4′-Dimethoxyphenyl)-1-(3″-pyridinyl)acrylonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O2
CAS Number:
Molecular Weight:
266.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98%

form

solid

potency

IC50 (3 micromolar for inhibition of EGF-receptor.)

mp

114-115 °C

solubility

DMSO or ethanol: soluble

storage temp.

2-8°C

SMILES string

COc1ccc(cc1OC)\C=C(\C#N)c2cccnc2

InChI

1S/C16H14N2O2/c1-19-15-6-5-12(9-16(15)20-2)8-14(10-17)13-4-3-7-18-11-13/h3-9,11H,1-2H3

InChI key

DBGZNJVTHYFQJI-UHFFFAOYSA-N

Biochem/physiol Actions

Non-phenolic tyrphostin-class tyrosine kinase inhibitor.

Preparation Note

Tyrphostin RG 13022 is known to suppress EGF-induced cancer cell proliferation under in vitro conditions and can also inhibit the growth of tumors in nude mice.
Tyrphostin RG 13022 is soluble in DMSO and ethanol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L F Gulli et al.
Cell growth & differentiation : the molecular biology journal of the American Association for Cancer Research, 7(2), 173-178 (1996-02-01)
A431 cells overexpress epidermal growth factor receptors (EGF-Rs) and are inhibited by EGF. We show that treatment of A431 cells with 10 nM EGF induced a 15-fold increase in EGF-R autophosphorylation, leading to inhibition of cell proliferation and morphological features
W Xian et al.
Cell growth & differentiation : the molecular biology journal of the American Association for Cancer Research, 6(11), 1447-1455 (1995-11-01)
The present study was designed to further investigate the role of the epidermal growth factor receptor (EGFr) in mouse skin tumor promotion by evaluating the status of the EGFr in tumor promoter-treated mouse epidermis and in mouse skin tumors. Female
B S Kondapaka et al.
Molecular and cellular endocrinology, 117(1), 53-58 (1996-03-01)
In prostate cancer cells, the binding of peptide growth factors to specific receptors increases tyrosine kinases (TK) activity to regulate cell proliferation, cell differentiation, and signaling processes. To determine whether inhibition of receptor TK activity inhibits tumor growth, we studied
K B Reddy et al.
Cancer research, 52(13), 3636-3641 (1992-07-01)
Human breast cancer cell proliferation is regulated by growth factors that bind to receptors with intrinsic tyrosine kinase (TK) activity, including the epidermal growth factor (EGF) receptor. To determine whether inhibition of receptor TK activity inhibits tumor growth, we studied
H L McLeod et al.
British journal of cancer, 74(11), 1714-1718 (1996-12-01)
Amplification and increased expression of many growth factor receptors, including the epidermal growth factor receptor (EGFR), has been observed in human tumours. One therapeutic strategy for overcoming EGF autocrine control of tumour growth is inhibition of EGFR protein tyrosine kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service