Skip to Content
Merck
All Photos(2)

Key Documents

T6375

Sigma-Aldrich

1-Thio-β-D-glucose sodium salt

Synonym(s):

β-D-Thioglucose, 1-β-D-Glucosylthiose, Glucothiose

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$221.00

$221.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$221.00

About This Item

Linear Formula:
C6H11O5SNa
CAS Number:
Molecular Weight:
218.20
EC Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.79

$221.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

yellow to tan

mp

130 °C (decomposes on heating)

solubility

water: 50 mg/mL, colorless to dark yellow

shipped in

wet ice

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](CO)O[C@@H](S[Na])[C@H](O)[C@H]1O

InChI

1S/C6H12O5S.Na.H/c7-1-2-3(8)4(9)5(10)6(12)11-2;;/h2-10,12H,1H2;;

InChI key

BJZCRDSEUWQHLR-UHFFFAOYSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service