Skip to Content
Merck
All Photos(2)

Key Documents

T5785

Sigma-Aldrich

2,4,5-Trichlorophenoxyacetic acid

BioReagent, suitable for plant cell culture, ≥95%, crystalline

Synonym(s):

Trioxone, 2,4,5-T

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OCH2CO2H
CAS Number:
Molecular Weight:
255.48
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

biological source

synthetic (organic)

grade

reagent

product line

BioReagent

Assay

≥95%

form

crystalline

technique(s)

cell culture | plant: suitable

color

off-white to yellow

mp

154-158 °C (lit.)

solubility

5% ethanol: 50, clear to slightly hazy, colorless to faintly yellow

application(s)

agriculture

SMILES string

OC(=O)COc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C8H5Cl3O3/c9-4-1-6(11)7(2-5(4)10)14-3-8(12)13/h1-2H,3H2,(H,12,13)

InChI key

SMYMJHWAQXWPDB-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

2,4,5-Trichlorophenoxyacetic acid has been used in the preparation of basal medium for leaflet explants.[1]

Biochem/physiol Actions

2,4,5-Trichlorophenoxyacetic acid (2,4,5-T) is a synthetic plant hormone (auxin) that promotes processes such as somatic embryogenesis and selective gene transcription. It is also a herbicide that defoliates broad-leafed plants. 2,4,5-T together with 2,4-D comprises the defoliant Agent Orange.

Preparation Note

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

G Hagen et al.
Molecular and cellular biology, 5(6), 1197-1203 (1985-06-01)
Nuclei isolated from excised soybean plumules that were treated with 2,4-dichlorophenoxyacetic acid (2,4-D) were active in transcription of four auxin-regulated genes or DNA sequences, which have been described previously (G. Hagen, A. Kleinschmidt, and T. Guilfoyle, Planta 162:147-153, 1984). The
J P Jouanneau et al.
Plant physiology, 96(2), 459-466 (1991-06-01)
When auxin was omitted during either the preparation or the culture of tobacco mesophyll protoplasts, as well as during both periods, synthesis of beta-glucanase was spontaneously induced. In contrast, when protoplasts were prepared and cultured in the presence of 16
The Effect of 2, 4, 5-T on Embryogenic Tissue Formation in Rose
Estabrooks T, et al.
Flavours and Fragrances (2008)
A M Wielandt et al.
The Biochemical journal, 341 ( Pt 1), 105-111 (1999-06-23)
The canalicular multispecific organic anion transporter, cMoat, is an ATP-binding-cassette protein expressed in the canalicular domain of hepatocytes. In addition to the transport of endo- and xenobiotics, cMoat has also been proposed to transport GSH into bile, the major driving
Anh Duc Ngo et al.
European journal of epidemiology, 25(1), 37-44 (2009-11-07)
The objective of this study is to conduct a meta-analysis of published and unpublished studies that examine the association between Agent Orange (AO) exposure and the risk of spina bifida. Relevant studies were identified through a computerized literature search of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service