Skip to Content
Merck
All Photos(1)

Key Documents

T4951

Sigma-Aldrich

2-((3-Trifluoromethyl)phenyl)histamine dimaleate

≥98% (HPLC), powder

Synonym(s):

(2-[2-(3-Trifluoromethyl)phenyl)-1H-imidazol-4-yl]ethanamine dimaleate, Compound 39

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$207.00

$207.00


Estimated to ship onApril 05, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$207.00

About This Item

Empirical Formula (Hill Notation):
C20H20F3N3O8
CAS Number:
Molecular Weight:
487.38
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

$207.00


Estimated to ship onApril 05, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

H2O: ~20 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.OC(=O)\C=C/C(O)=O.NCCc1c[nH]c(n1)-c2cccc(c2)C(F)(F)F

InChI

1S/C12H12F3N3.2C4H4O4/c13-12(14,15)9-3-1-2-8(6-9)11-17-7-10(18-11)4-5-16;2*5-3(6)1-2-4(7)8/h1-3,6-7H,4-5,16H2,(H,17,18);2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-

InChI key

GGDUVPKRZGATLR-SPIKMXEPSA-N

Application

2-((3-Trifluoromethyl)phenyl)histamine dimaleate has been used to study the induction of goblet cell secretion in rats3. 2-((3-Trifluoromethyl)phenyl)histamine dimaleate has also been used to study the role of H1 receptor in COX-2 regulation and prostanoid homeostasis in human vascular endothelial cells4.

Biochem/physiol Actions

2-((3-Trifluoromethyl)phenyl)histamine dimaleate was found to be the most potent and selective H1 histamine receptor agonist of a panel of compounds in functional in vitro studies on histamine H2, H3, and other neurotransmitter receptors. It showed better potency at the guinea pig H1 histamine receptor than at the human H1 histamine receptor (pKi : 5.9). The selectivity of 2-((3-Trifluoromethyl)phenyl)histamine dimaleate was found to be 2138 (H1:H2), > 64 (H1:H3), 1000 (H1:M3), 105 (H1: a1), 708 (H11), and 71 (H1:5HT2A). 2-((3-Trifluoromethyl)phenyl)histamine dimaleate does not cross the blood-brain barrier.
Potent and selective H1 histamine receptor agonist.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

2-((3-Trifluoromethyl)phenyl)histamine dimaleate is soluble in water at approximately 20 mg/ml.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roland Seifert et al.
The Journal of pharmacology and experimental therapeutics, 305(3), 1104-1115 (2003-03-11)
Species isoforms of histamine H2-, H3-, and H4-receptors differ in their pharmacological properties. The study aim was to dissect differences between the human H1R (hH1R) and guinea pig H1R (ghH1R). We coexpressed hH1R and gpH1R with regulators of G-protein signaling
C Leschke et al.
Journal of medicinal chemistry, 38(8), 1287-1294 (1995-04-14)
New histamine derivatives characterized by a (substituted) aryl, heteroaryl, benzyl, or heteroarylmethyl substituent in the C2 position of the imidazole ring have been prepared from appropriate imidates or amidines, respectively, and 2-oxo-4-phthalimido-1-butyl acetate (1). The compounds were screened as potential
Densen Hayashi et al.
Investigative ophthalmology & visual science, 53(6), 2993-3003 (2012-04-03)
The purpose of this study was to determine the effect of histamine and its receptors on goblet cell secretion. Cultured rat and human goblet cells were grown in RPMI 1640. Goblet cell secretion of high molecular weight glycoconjugate was measured
Xiaoyu Tan et al.
Journal of immunology (Baltimore, Md. : 1950), 179(11), 7899-7906 (2007-11-21)
Although histamine plays an essential role in inflammation, its influence on cyclooxygenases (COX) and prostanoid homeostasis is not well understood. In this study, we investigated the effects of histamine on the expression of COX-1 and COX-2 and determined their contribution
Trinidad Raices et al.
Journal of molecular endocrinology, 63(3), 187-197 (2019-08-16)
Testicular Leydig cells (LC) are modulated by several pathways, one of them being the histaminergic system. Heme oxygenase-1 (HO-1), whose upregulation comprises the primary response to oxidative noxae, has a central homeostatic role and might dysregulate LC functions when induced.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service