Key Documents
T4793
Nalgene® centrifuge tubes, Round Style 3117
nominal capacity 100 mL
About This Item
UNSPSC Code:
41121703
material
lipped
polycarbonate
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10
manufacturer/tradename
Nalgene®
nominal capacity
100 mL
General description
These high-speed polycarbonate tubes withstand temperatures from -135° to +135°C (-211° to +275°F)
  • Designed with round bottoms to fit high-speed rotors
  • Molding technique produces uniform wall thickness for increased strength
  • Uniform diameter (no taper) for better rotor fit and force tolerance
  • Lip allows easier retrieval from rotor cavities
  • Economical choice - may also be used as test tubes
  • Longer-lasting than polycarbonate tubes under conditions of repeated autoclaving
  • Autoclavable/Transparent
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)