Merck
All Photos(5)
Key Documents
T4162
Tweezers
Style #3C, very sharp, non-magnetic alloy (tips may be restraightened when bent)
Synonym(s):
fine forceps, fine tweezers, needle tweezers
Select a Size
1 EA
$284.00
$284.00
Estimated to ship onMay 24, 2025
Select a Size
Change View
1 EA
$284.00
About This Item
UNSPSC Code:
27112105
NACRES:
MA.01
$284.00
Estimated to ship onMay 24, 2025
material
non-magnetic alloy (tips may be restraightened when bent)
packaging
pack of 1 ea
manufacturer/tradename
Ted Pella 523-C
size
110 mm
General description
Specifically designed for microscopy applications. All are ultra-high precision tools of Swiss manufacture.
Choose from one of the most recent versions:
Certificates of Analysis (COA)