Skip to Content
Merck
All Photos(1)

Key Documents

T2455

Sigma-Aldrich

T3MG

Synonym(s):

(±)-threo-3-Methylglutamic acid, 3MG, threo-3-Methylglutamate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$211.00
25 MG
$682.00

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$211.00
25 MG
$682.00

About This Item

Empirical Formula (Hill Notation):
C6H11NO4
CAS Number:
Molecular Weight:
161.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

form

powder

color

white to off-white

solubility

H2O: >1.5 mg/dL

suitability

suitable for TLC (single spot)

storage temp.

room temp

SMILES string

C[C@@H](CC(O)=O)[C@@H](N)C(O)=O

InChI

1S/C6H11NO4/c1-3(2-4(8)9)5(7)6(10)11/h3,5H,2,7H2,1H3,(H,8,9)(H,10,11)/t3-,5+/m0/s1

InChI key

FHJNAFIJPFGZRI-WVZVXSGGSA-N

Biochem/physiol Actions

T3MG is a selective inhibitor of excitatory amino acid transporters GLT-1, EAAT2, and EAAT4.
T3MG is a selective inhibitor of excitatory amino acid transporters GLT-1, EAAT2, and EAAT4. T3MG has long been utilized as a gold standard in the study of general EAAT function. It has more recently been shown to be selective for EAAT2 and EAAT4 as compared to EAAT1 and EAAT3 in electrophysiology and glutamate/aspartate uptake assays. Although there are other EAAT2-selective compounds, this is the first tool with selectivity for EAAT4. T3MG is not a substrate for the transporters themselves, nor does it have activity at glutamate ion channels.

Features and Benefits

This compound is featured on the Excitatory Amino Acid Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

T3MG dissolves in water at a concentration that is greater than 1.5 mg/ml.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lois Slator et al.
Journal of aerosol medicine and pulmonary drug delivery, 27 Suppl 1, S37-S43 (2014-01-31)
Valved holding chambers (VHCs) are accessory devices for pressurized metered dose inhalers (pMDIs). Use of a VHC may help overcome coordination issues associated with drug delivery via the pMDI alone. Previous work has established that aspects of VHC use, including
Guide to Receptors and Channels (GRAC), 4th Edition.
British journal of pharmacology, 158 Suppl 1, S1-254 (2009-11-04)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service