Key Documents
T2168
Sigma-Aldrich
Nalgene® centrifuge tubes, Oak Ridge Style 3114
capacity 10 mL, pack of 2 ea
Synonym(s):
High-Speed ​​Centrifuge Tubes
Select a Size
2 EA
$187.00
$187.00
PRICE WITHOUT NATIONAL TAXES
Available to ship onApril 07, 2025Details
Select a Size
Change View
2 EA
$187.00
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
$187.00
PRICE WITHOUT NATIONAL TAXES
Available to ship onApril 07, 2025Details
material
FEP
Tefzel® closure
round bottom tube
screw top cap
sterility
non-sterile
feature
RCF 10,000 × g
packaging
pack of 2 ea
manufacturer/tradename
Nalgene® 3114-0010
capacity
10 mL
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)