Skip to Content
Merck
All Photos(1)

Key Documents

T2000

Sigma-Aldrich

Testosterone β-D-glucuronide

Synonym(s):

17β-Hydroxy-4-androsten-3-one 17-D-glucuronide, 4-Androsten-17β-ol-3-one 17-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H36O8
CAS Number:
Molecular Weight:
464.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

CC12CCC3C(CCC4=CC(=O)CCC34C)C1CCC2OC5OC(C(O)C(O)C5O)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Application

Testosterone has been used as a substrate for UDP-glucuronyltransferase based drug metabolism assays using microsome-encapsulation techniques[1].

Biochem/physiol Actions

Androgens direct the development of the male phenotype during embryogenesis and at puberty. Testosterone is an androgen that is secreted by the testis. This hormone is converted to dihydrotestosterone in the target tissues where it appears to mediate several biological functions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Di Luigi et al.
Journal of endocrinological investigation, 32(5), 445-453 (2009-05-23)
To describe serum and urinary hormones, androgens metabolites and testosterone/epitestosterone ratio profiles after testosterone administration in male hypogonadal volunteers, and to evaluate their possible usefulness in detecting doping with testosterone in treated hypogonadal athletes. Controlled open label design vs placebo;
L Dehennin et al.
European journal of endocrinology, 130(1), 53-59 (1994-01-01)
In this cross-sectional study on 140 subjects, several testosterone and epitestosterone metabolites have been analyzed by gas chromatography-mass spectrometry associated with stable isotope dilution, a technique requested for doping analysis in general, and detection of exogenous testosterone supply in particular.
E Palonek et al.
The Journal of steroid biochemistry and molecular biology, 55(1), 121-127 (1995-10-01)
In an attempt to find optimal markers of exogenous testosterone (T) administration in male athletes, a number of compounds were measured in 11 healthy men before and after 3, 6 and 9 months of weekly administration of 250 mg i.m.
Genetic variations in UDP-glucuronosyl transferase 2B17: implications for testosterone excretion profiling and doping control programs.
Borislav Starcevic et al.
Clinical chemistry, 54(12), 1945-1947 (2008-10-18)
L Dehennin
The Journal of endocrinology, 142(2), 353-360 (1994-08-01)
Testosterone administration to male athletes can be safely detected in the vast majority of cases by the urinary excretion ratio of testosterone to epitestosterone glucuronides (TG/EG), which may not exceed 6. Some rare cases of physiologically high TG/EG ratios (between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service