Skip to Content
Merck
All Photos(2)

Key Documents

T1894

Sigma-Aldrich

cis-9-Tetradecenol

~95%

Synonym(s):

(Z)-9-TDOL, Myristoleyl alcohol, Z-9-TDOL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CH(CH2)8OH
CAS Number:
Molecular Weight:
212.37
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

~95%

refractive index

n20/D 1.457 (lit.)

storage temp.

−20°C

SMILES string

[H]\C(CCCC)=C(/[H])CCCCCCCCO

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pheromone for Homoeosoma electellum

Packaging

Sealed ampule (liquid).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S B Jiang et al.
Journal of immunological methods, 126(1), 29-37 (1990-01-24)
Perforin-mediated lysis consists of at least three steps: perforin binding to the target cell, insertion into the plasma membrane, and polymerization to form pores. Perforin binding, the first step, is critical for pore formation. Accordingly, a competition assay was here
T J O'Leary et al.
Biophysical journal, 50(6), 1053-1059 (1986-12-01)
The effects of cis- and trans-9,10-tetradecenols on the phase transitions of dimyristoyl-, dipalmitoyl-, and distearoyl-phosphatidylcholines were investigated using high sensitivity scanning calorimetry and Raman spectroscopy. Both alcohols lowered the gel to liquid crystalline phase transition temperatures for all three phosphatidylcholines
Erik van Bergen et al.
Proceedings. Biological sciences, 280(1758), 20130102-20130102 (2013-03-08)
Inbreeding depression results from mating among genetically related individuals and impairs reproductive success. The decrease in male mating success is usually attributed to an impact on multiple fitness-related traits that reduce the general condition of inbred males. Here, we find
D Morse et al.
The Journal of biological chemistry, 259(1), 475-480 (1984-01-10)
Highly sensitive, luminescent assays have been developed to measure enzyme activities involved in the metabolism of a major class of insect pheromones which includes fatty aldehydes, alcohols, and their acetate esters. These assays have been applied to measure the in
D M Ojcius et al.
Molecular immunology, 27(9), 839-845 (1990-09-01)
Cytotoxic T lymphocytes (CTL) release from their granules a 70 kDa protein, called PFP, perforin or cytolysin, which inserts into the target cell plasma membrane in its monomeric form. Here it polymerizes into a macromolecular complex forming pores as large

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service