Skip to Content
Merck
All Photos(1)

Key Documents

T1819

Sigma-Aldrich

Trizma® hydrochloride solution

Trizma® hydrochloride solution
1 of 1 reviewers received a sample product or took part in a promotion

pH 7.0, BioPerformance Certified, 1 M, suitable for cell culture

Synonym(s):

Tris hydrochloride solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$60.80
1 L
$115.00

$60.80


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$60.80
1 L
$115.00

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

$60.80


Estimated to ship onMay 30, 2025


Request a Bulk Order

grade

BioPerformance Certified
Molecular Biology
for molecular biology

form

liquid

concentration

1 M

technique(s)

cell culture | mammalian: suitable

impurities

DNase, RNase, Protease, none detected
bioburden, tested
endotoxin, tested
≤5 ppm Heavy metals (as Pb)

pH

7.0

useful pH range

7.0-9.0

absorption

≤0.05 at 290 at 40%

Looking for similar products? Visit Product Comparison Guide

General description

A series of Pre-mixed solutions of TRIZMA Base and TRIZMA HCl to provide commonly used pH values for Tris buffers. No mixing or pH adjustment necessary. Guaranteed accuracy ± 0.1 pH units.
Tris(hydroxymethyl)aminomethane (Tris) is a standard buffer in biological sciences and has a pH buffering range between 7.0 - 9.0.[1] It is applicable in electrophoretic studies and capillary electrochromatography.[2]

Application

Trizma® hydrochloride solution has been used:
  • as a buffer in reverse transcription and DNase I treatment reactions for RNA isolated from Toxoplasma gondii[3]
  • in the preparation of tris- ethylenediaminetetraacetic acid (EDTA) (TE) buffer for bacterial pellet resuspension for plasmid isolation studies[4]
  • for polylactic acid nanoparticle resuspension, storage, and pH-based stability studies[5]

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A review of acellular immersion tests on bioactive glasses-influence of medium on ion release and apatite formation
Nommeots-Nomm A, et al.
International Journal of Applied Glass Science, 11(3) (11)
(Un) suitability of the use of pH buffers in biological, biochemical and environmental studies and their interaction with metal ions-a review
Ferreira CMH, et al.
Royal Society of Chemistry Advances, 5(39) (5)
Junya Yamagish et al.
Methods in molecular biology (Clifton, N.J.), 1201, 193-205 (2014-11-13)
Parasites change their transcriptional systems in different developmental stages and in response to environmental changes. To investigate the molecular mechanisms that underlie transcriptional regulation, it is essential to identify the exact positions of the transcriptional start sites (TSSs) and characterize
Production and characterisation of biodegradable PLA nanoparticles loaded with thymol to improve its antimicrobial effect
Marcet I, et al.
Journal of Food Engineering, 239 (239)
Kaichun Zhu et al.
Nature protocols, 1(6), 3088-3093 (2007-04-05)
This protocol describes a streamlined method of plasmid DNA extraction by continual thermal lysis, a modification of the basic boiling lysis technique, to simplify the processing of large volumes of Escherichia coli cultures. Fermented bacteria are harvested using a hollow

Questions

  1. What are the storage conditions for this product?

    1 answer
    1. This product is to be stored in room temperature. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

      For more safety and handling information, please view the product Safety Data Sheet:
      https://www.sigmaaldrich.com/sds/sigma/t1819

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

    • Review 1
    • Votes 0
    4 out of 5 stars.

    NaOCl Sodium Hypochlorite

    All good, however, the delivery time was longer than expected.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service