Tumor Necrosis Factor-α (TNF-α) also refers as cachectin is a cytokine produced in response to immunological challenges such as bacteria (lipopolysaccharides), parasites, mitogens, viruses and other cytokines. It has essential roles in antitumor activity, inflammation, septic shock, immune modulation, anorexia, cachexia, hematopoiesis and viral replication. It also facilitates the growth of human fibroblasts, collagenase production as well as can activates polymorphonuclear neutrophils and osteoclasts. Anti-Tumor Necrosis Factor-α antibody can be used in neutralization and immunoblotting (working concentration-0.1-0.2 μg/ml) for detecting porcine TNF-α. Goat anti-Tumor Necrosis Factor-α antibody reacts specifically with recombinant porcine tumor necrosis factor α.
Specificity
The antibody neutralizes the biological activity of recombinant porcine TNF-α.
Immunogen
recombinant porcine TNF-α expressed in Escherichia coli.
Application
Anti-Tumor Necrosis Factor-α antibody can be used in immunohistochemistry and indirect ELISA.
Physical form
Lyophilized from a 0.2 μm filtered solution of phosphate buffered saline.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
When leukocytes are exposed to mitogens or antigens in vitro, they release bone-resorbing activity into the culture supernatants which can be detected by bioassay. Like many lymphocyte-monocyte products, this activity has been difficult to purify because of its low abundance
The Journal of experimental medicine, 162(6), 2163-2168 (1985-12-01)
Cachectin/TNF (tumor necrosis factor), an endotoxin-induced murine macrophage hormone implicated in the pathogenesis of cachexia and shock, has been found capable of stimulating collagenase and prostaglandin E2 (PGE2) production by isolated human synovial cells and dermal fibroblasts. This bioactivity associated
Molecular medicine reports, 13(2), 1250-1256 (2015-12-10)
Complement anaphylatoxins have been investigated extensively; however, the role of complement anaphylatoxin C4a in hyperoxic lung injury has yet to be investigated. To the best of our knowledge, the present study is the first to demonstrate the role of C4a
Science (New York, N.Y.), 230(4728), 943-945 (1985-11-22)
Modulation of the growth of human and murine cell lines in vitro by recombinant human tumor necrosis factor-alpha (rTNF-alpha) and recombinant human interferon-gamma (rIFN-gamma) was investigated. rTNF-alpha had cytostatic or cytolytic effects on only some tumor cell lines. When administered
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.