Skip to Content
Merck
All Photos(1)

Key Documents

T180

Sigma-Aldrich

NK2 Tachykinin Receptor human

recombinant, expressed in CHO cells, membrane suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

recombinant

expressed in CHO cells

Quality Level

form

membrane suspension

packaging

vial of 50 units

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TACR2(6865)

Physical form

Supplied in 50 mM Tris-HCl, pH 7.4, containing 10% glycerol and 1% BSA. Actual concentrations and specific binding capacity are provided with each lot.

Analysis Note

Recommended assay conditions are supplied on the product data sheet. This is a bioassayed product.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Nakajima et al.
The Journal of biological chemistry, 267(4), 2437-2442 (1992-02-05)
The mammalian tachykinin system consists of three distinct peptides, substance P, substance K, and neuromedin K, and possesses three corresponding receptors. In this investigation we examined intracellular signal transduction of the individual tachykinin receptors by transfection and stable expression of
Daolin Tang et al.
American journal of respiratory cell and molecular biology, 41(6), 651-660 (2009-03-07)
The pathogenesis of sepsis is mediated in part by the pathogen-associated molecular pattern molecule bacterial endotoxin, which stimulates macrophages to sequentially release early (e.g., TNF-alpha, IL-1beta) and late (e.g., high-mobility group box [HMGB] 1 protein) proinflammatory mediators. The recent discovery
Long Sun et al.
Molecular medicine reports, 17(2), 3274-3280 (2017-12-20)
The purpose of the present study was to examine the effects of myricetin on reducing cerebral ischemia injury in a rat model. A rat model of permanent middle cerebral artery occlusion (pMCAO) was used in the present study. Rats were
Benjamin R Pryce et al.
Biochimica et biophysica acta. Molecular cell research, 1868(2), 118917-118917 (2020-12-02)
Duchenne's muscular dystrophy (DMD) is a severe muscle wasting disorder characterized by the loss of dystrophin expression, muscle necrosis, inflammation and fibrosis. Ongoing muscle regeneration is impaired by persistent cytokine stress, further decreasing muscle function. Patients with DMD rarely survive
Haiyoung Jung et al.
The Journal of biological chemistry, 283(50), 34541-34553 (2008-10-25)
Murine protein serine/threonine kinase 38 (MPK38) is a member of the AMP-activated protein kinase-related serine/threonine kinase family that plays an important role in various cellular processes, including cell cycle, signaling pathways, and self-renewal of stem cells. Here we demonstrate a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service