Skip to Content
Merck
All Photos(1)

Key Documents

T1581

Sigma-Aldrich

TLQP-21 (mouse, rat)

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$522.00
1 MG
$927.00

$522.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

Select a Size

Change View
500 μG
$522.00
1 MG
$927.00

About This Item

Linear Formula:
H-Thr-Leu-Gln-Pro-Pro-Ala-Ser-Ser-Arg-Arg-Arg-His-Phe-His-His-Ala-Leu-Pro-Pro-Ala-Arg-OH
Molecular Weight:
2432.75
UNSPSC Code:
12352200
NACRES:
NA.32

$522.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

shipped in

wet ice

Quality Level

storage temp.

−20°C

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Young Kim et al.
The Journal of biological chemistry, 295(48), 16328-16341 (2020-09-06)
Acute kidney injury (AKI) is a common clinical condition associated with diverse etiologies and abrupt loss of renal function. In patients with sepsis, rhabdomyolysis, cancer, and cardiovascular disorders, the underlying disease or associated therapeutic interventions can cause hypoxia, cytotoxicity, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service