Skip to Content
Merck
All Photos(1)

Key Documents

T1016

Sigma-Aldrich

Trichlormethiazide

≥98%

Synonym(s):

6-Chloro-3-(dichloromethyl)-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide
(+/-)-, NSC 61560

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8Cl3N3O4S2
CAS Number:
Molecular Weight:
380.66
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98%

SMILES string

NS(=O)(=O)c1cc2c(NC(NS2(=O)=O)C(Cl)Cl)cc1Cl

InChI

1S/C8H8Cl3N3O4S2/c9-3-1-4-6(2-5(3)19(12,15)16)20(17,18)14-8(13-4)7(10)11/h1-2,7-8,13-14H,(H2,12,15,16)

InChI key

LMJSLTNSBFUCMU-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Uzu et al.
Journal of hypertension, 23(4), 861-865 (2005-03-19)
We examined whether thiazide diuretics could restore nocturnal blood pressure (BP) decline and reduce urinary protein excretion in patients with glomerulopathy treated with angiotensin II modulators (angiotensin-converting enzyme inhibitors or angiotensin II receptor blockers). Twenty-five Japanese outpatients (11 men, 14
Kohei Ueda et al.
Internal medicine (Tokyo, Japan), 51(12), 1549-1553 (2012-06-26)
Gitelman's syndrome (GS), an inherited disorder due to loss of function of ion channels and transporters such as Na-Cl co-transporter (NCCT) in distal convoluted tubules, is characterized by hypokalemia, hypomagnesemia, hypocalciuria, metabolic alkalosis and hyperreninemic-hyperaldosteronism. A 39-year-old man was admitted
Kenichiro Miura et al.
Pediatric nephrology (Berlin, Germany), 24(6), 1235-1238 (2009-01-21)
The administration of angiotensin-converting enzyme inhibitors (ACEIs) or angiotensin type 1 receptor blockers (ARBs) to pregnant women has been reported to cause ACEI/ARB fetopathy, including oligohydramios, pulmonary hypoplasia, renal insufficiency, limb contracture, and fetal hypotension in the child. Most of
Susumu Ogawa et al.
Hypertension (Dallas, Tex. : 1979), 47(4), 699-705 (2006-03-01)
We tested the hypothesis that blockade of angiotensin II type 1 receptors reduces oxidative stress markers in parallel with urinary albumin and type IV collagen excretions. Sixty-six diabetic patients with nephropathy were randomly assigned to either the angiotensin II receptor
I Kuwajima et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 24(5), 475-480 (2001-10-26)
A randomized prospective controlled study, the National Interventional Cooperative Study in Elderly Hypertensives (NICS-EH), previously demonstrated that the preventive effect of the long-acting calcium channel blocker nicardipine on the cardiovascular endpoint was similar to that of the diuretic, trichlormethiazide. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service