Skip to Content
Merck
All Photos(1)

Documents

T0956

Sigma-Aldrich

Sigma Universal Transfection Reagent

Polymer reagent for transfecting common mammalian cell lines

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

grade

for molecular biology

Quality Level

sterility

aseptically filled

form

liquid (buffered aqueous solution)

usage

sufficient for 110 transfections (3.5 cm dish)
sufficient for 40 transfections (10 cm dish)
sufficient for 65 transfections (6 cm dish)

concentration

2 mg/mL

storage temp.

−20°C

General description

Sigma Universal Transfection Reagent is a unique formulation of a biodegradable, proprietary cationic polymer conjugated to a cationic lipid. Universal Transfection Reagent is used for transfection of nucleic acids into common eukaryotic cell lines. Fast and easy protocol is compatible with serum, serum-free medium and antibiotics. For most cells, medium change after transfection is not necessary.

Application

Suitable for transient and stable transfection of nucleic acids into cultured eukaryotic cells. Use approximately 1-3 μl Universal Transfection Reagent per μg DNA. Protocol optimization provides very efficient transfection. The following cells have been successfully transfected using Sigma Universal Transfection Reagent:

Excellent for:
BHK-21
CHO
CHOK1
HEK293
HeLa
NIH3T3

Good for:
3LL
6CSFME
ATT20B16-F0
B16-F10
BAEC
Ca Ski
COS-7
CV-1
D 407
HUVEC
L929
MCF-7
MDCK
Neuro2A
SAOS-2
SHEP
SiHa
SKOV3

Features and Benefits

  • Fast, easy protocol
  • Suitable for stable and transient transfection
  • High efficiency for a wide variety of cell types including primary cells
  • Compatible with both serum and serum-free transfection protocols

Components

Note the formulation of Sigma Universal Transfection Reagent is confidential.

Principle

Sigma Universal Transfection Reagent is a cationic polymer conjugated to a cationic lipid. When mixed with plasmid DNA, the reagent forms stable complexes that can be directly added to the cell culture. The cells take up the complexes by endocytosis, the polymer is cleaved from the complex, and the plasmid DNA is quickly released into the cytoplasm.

Other Notes

Complex formation is inhibited by serum, but once stable complexes have formed, the presence of serum lowers the toxicity of transfection.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pan Pan et al.
Nature communications, 12(1), 4664-4664 (2021-08-04)
Excessive inflammatory responses induced upon SARS-CoV-2 infection are associated with severe symptoms of COVID-19. Inflammasomes activated in response to SARS-CoV-2 infection are also associated with COVID-19 severity. Here, we show a distinct mechanism by which SARS-CoV-2 N protein promotes NLRP3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service