STS0033
Agarose tablets
Low EEO, For molecular biology (200 x 500mg tablets)
Synonym(s):
3,6-Anhydro-α-L-galacto-β-D-galactan, Agarose LE, Agarose tablets
About This Item
Recommended Products
biological source
algae (marine)
grade
Molecular Biology
for molecular biology
form
tablet
impurities
≤10.0% moisture content
color
white
EEO
0.09-0.13 (-Mr)
transition temp
transition temp 34.5-37.5 °C
gel strength
≥1200 g/cm2 (1% gel)
anion traces
sulfate (SO42-): ≤0.15%
suitability
suitable for electrophoresis
suitable for molecular biology
foreign activity
DNase, RNase, none detected
SMILES string
O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O
InChI
1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1
InChI key
MJQHZNBUODTQTK-WKGBVCLCSA-N
General description
Application
- Separation and analysis of nucleic acids by agarose gel electrophoresis, Northern Blot, or Southern Blot[2]
- Single Cell Gel Electrophoresis (SCGE), also known as comet assays
- Hydrocolloid gels to study the structural influence of gels on the release of carbohydrates[3]
- Ouchterlony double immunodiffusion plates used to quantify antibodies and antigens
- Radial immunodiffusion (RID)
Features and Benefits
- Qualified for molecular biology applications
- Free of endonucleases: DNase and RNase
- Convenient, quick and easy to use
- Low EEO and gelling temperature for routine analysis
- High gel strength 1,200 g/cm2 (1% gel)
- Separates DNA & RNA fragments 100bp - 23kb in 2% - 0.5% agarose gel, respectively.
- Northern or Southern blotting, preferred for fragment sizes >1kb
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service