Skip to Content
Merck
All Photos(1)

Key Documents

SRP8008

Sigma-Aldrich

FTO mouse

recombinant, expressed in E. coli, His tagged, >90% (SDS-PAGE)

Synonym(s):

EC 1.14.11, FATSO, Fat mass and obesity-associated protein, Preadipocyte factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$462.00

$462.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
$462.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$462.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

mouse

recombinant

expressed in E. coli

tag

His tagged

Assay

>90% (SDS-PAGE)

form

liquid

mol wt

~55 kDa by SDS-PAGE

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

≥0.2 mg/mL

impurities

<1 EU/μg endotoxin, tested

color

clear

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Fto(26383)

General description

FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ′fused toes′ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.

Physical form

Solution in 55 mM TRIS-HCl, pH 8.2, containing 150 mM NaCl.

Other Notes

Mouse FTO (aa 2-502) is fused at the N-terminus to a His-tag.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service