Skip to Content
Merck
All Photos(2)

Key Documents

SRP6560

Sigma-Aldrich

Wnt-2 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

INT1L1, IRP, Int-1-like protein 1, Int-1-related protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

lyophilized

mol wt

39.9 kDa

packaging

pkg of 1 mg
pkg of 10 μg
pkg of 50 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... WNT2(7472)

Biochem/physiol Actions

Wnt gene family members, including Wnt-1 through Wnt-10, play a key role in regulating cellular growth and differentiation. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis.[1] Wnt family member-2 (Wnt-2) has a role in activating β-catenin.[2] It has been shown to have a role in cardiogenesis.[3] The protein is overexpressed in tumors and cancers.[4][1]

Physical form

Lyophilized without additives.

Reconstitution

Reconstitute in sterile ddH2O to a concentration ≥ 100 μg/mL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wnt2 complements Wnt/?-catenin signaling in colorectal cancer
Youn-Sang Jung
Oncotarget, 6(35), 37257-37268 (2015)
Wnt2 as a new therapeutic target in malignant pleural mesothelioma
Julien Mazieres
International Journal of Cancer. Journal International Du Cancer (2005)
Steven Hyman
Autism: The Science of Mental Health (2013)
Overexpression of Wnt-2 in colorectal cancers.
Park JK
Neoplasma, 56(2), 119-123 (2009)
Wnt signal transduction pathways
Yuko Komiya
Organogenesis, 4(2), 68-75 (2008)

Articles

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service