Skip to Content
Merck
All Photos(1)

Key Documents

SRP6549

Sigma-Aldrich

Alkaline Phosphatase from calf intestine

≥98% (SDS-PAGE)

Synonym(s):

ALP, ALP-1, ALPP, ALPPL, ALPPL2, PALP, PLAP, PLAP-1

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352205

biological source

bovine (calf) intestine

Assay

≥98% (SDS-PAGE)

form

freeze-dried protein glass

packaging

pkg of 1 mg
pkg of 10 μg
pkg of 50 μg

impurities

3000-6,000 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

bovine (calf) ... ALPL(280994)

General description

Alkaline phosphatase (ALP) is a hydrolase enzyme. It is a highly active form in calf intestinal mucosa.[1][2] The calf intestinal alkaline phosphatase is a dimeric glycoprotein with a molecular weight of about 140kDa.[3]

Application

Alkaline phosphatase from calf intestine has been used as a standard in alkaline phosphatase assay.[4]

Biochem/physiol Actions

Alkaline phosphatase (ALP) is responsible for removing phosphate groups at the 5- and 3- positions from many types of molecules, such as nucleotides, proteins, and alkaloids.[2] It can also modify lipopolysaccharides by dephosphorylation of the lipid A moiety, thereby making it less toxic than the unmodified lipid A.[5]

Physical form

Lyophilized without any additives.

Reconstitution

Reconstitute in distilled water or dilute buffer.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrophosphate inhibits mineralization of osteoblast cultures by binding to mineral, up-regulating osteopontin, and inhibiting alkaline phosphatase activity.
Addison WN
The Journal of Biological Chemistry, 282, 15872-15883 (2007)
Quantitation of surface coverage of oligonucleotides bound to chip surfaces: a fluorescence-based approach using alkaline phosphatase digestion.
Pal S
Lab on a chip, 8, 1332-1341 (2008)
Intestinal alkaline phosphatase detoxifies lipopolysaccharide and prevents inflammation in zebrafish in response to the gut microbiota.
Cell host & microbe, 2, 371-382 (2007)
Affinity-chromatography purification of alkaline phosphatase from calf intestine.
Brenna O
The Biochemical Journal, 151, 291-296 (1975)
Intestinal alkaline phosphatase. Physical properties and quaternary structure.
M Fosset et al.
Biochemistry, 13(9), 1783-1788 (1974-04-23)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service