Skip to Content
Merck
All Photos(1)

Documents

SRP6518

Sigma-Aldrich

Plasminogen from human plasma

≥95% (SDS-PAGE)

Synonym(s):

PLG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

90.0 – 92.0 kDa

packaging

pkg of 1 mg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PLG(5340)

General description

Plasminogen is the inactive precursor of plasmin. Plasminogen is activated by the action of either tissue plasminogen activator (tPA), which primarily activates the fibrinolytic (thrombolytic) activity of plasmin, or urokinase plasminogen activator (uPA), which is associated with extracellular matrix remodeling and cell migration.
Plasminogen is encoded by the gene mapped to human chromosome 6q26–27. The encoded protein is characterized with a leader, kringle and protease domains.

Biochem/physiol Actions

Plasmin dissolves the fibrin of blood clots and acts as a proteolytic factor in a variety of other processes including embryonic development, tissue remodeling, tumor invasion, and inflammation. It cleaves fibrin/fibrinogen and blood coagulation factors V/Va and VIII/VIIIa. It activates matrix metalloproteinases by cleaving the inactive proenzymes. Plasmin is also involved in the activation of some growth factors, such as vascular endothelial growth factor (VEGF) and transforming growth factor β (TGF-β).

Physical form

Lyophilized from 20 mM Tris-HCl, pH 7.5, with 2 mM EDTA.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matrix metalloproteinase-1 and -9 activation by plasmin regulates a novel endothelial cell-mediated mechanism of collagen gel contraction and capillary tube regression in three-dimensional collagen matrices.
Davis GE, et al.
Journal of Cell Science, 114, 917-930 (2001)
Genetic linkage between lipoprotein (a) phenotype and a DNA polymorphism in the plasminogen gene.
Drayna D T, et al.
Genomics, 3(3), 230-236 (1988)
Plasmin activates the lymphangiogenic growth factors VEGF-C and VEGF-D.
McColl BK, et al.
The Journal of Experimental Medicine, 198, 863-868 (2003)
Dante Disharoon et al.
Journal of thrombosis and haemostasis : JTH, 20(2), 486-497 (2021-12-10)
To reestablish blood flow in vessels occluded by clots, tissue plasminogen activator (tPA) can be used; however, its efficacy is limited by transport to and into a clot and by the depletion of its substrate, plasminogen. To overcome these rate
Minor Role of Plasminogen in Complement Activation on Cell Surfaces.
Hyvarinen S and Jokiranta TS
PLoS ONE, 10, e0143707-e0143707 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service