Skip to Content
Merck
All Photos(1)

Key Documents

SRP6083

Sigma-Aldrich

ALDH2 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

ALDH class 2, ALDH-E2, ALDHI, ALDM, Aldehyde dehydrogenase, mitochondrial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

54.5 kDa

packaging

pkg of 100 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ALDH2(217)

General description

ALDH2 (Aldehyde dehydrogenase 2 family) belongs to the aldehyde dehydrogenase family which catalyze the chemical transformation from acetaldehyde to acetic acid and is the second enzyme of the major oxidative pathway of alcohol metabolism. There are two major liver isoforms of this enzyme, cytosolic and mitochondrial, and they can be also distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. ALDH2 has been shown to interact with GroEL.
The ALDH2 gene is mapped to human chromosome 12q24.12.[1]

Biochem/physiol Actions

Aldehyde dehydrogenases catalyze the chemical transformation from acetaldehyde to acetic acid and ALDH2 (Aldehyde dehydrogenase 2 family) is the major enzyme of the oxidative pathway of alcohol metabolism.[2] Overexpression of ALDH2 inhibits cell injury induced by acetaldehyde and prevents activation of stress signals.[3]
It is actively involved in the metabolism of both endogenous and exogenous substances.[4]

Physical form

1 mg/mL solution in 20 mM Tris-HCl buffer (pH 7.5) containing 1 mM DTT, 1 mM EDTA and 10% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MSAAATQAVP APNQQPEVFC NQIFINNEWH DAVSRKTFPT VNPSTGEVIC QVAEGDKEDV DKAVKAARAA FQLGSPWRRM DASHRGRLLN RLADLIERDR TYLAALETLD NGKPYVISYL VDLDMVLKCL RYYAGWADKY HGKTIPIDGD FFSYTRHEPV GVCGQIIPWN FPLLMQAWKL GPALATGNVV VMKVAEQTPL TALYVANLIK EAGFPPGVVN IVPGFGPTAG AAIASHEDVD KVAFTGSTEI GRVIQVAAGS SNLKRVTLEL GGKSPNIIMS DADMDWAVEQ AHFALFFNQG QCCCAGSRTF VQEDIYDEFV ERSVARAKSR VVGNPFDSKT EQGPQVDETQ FKKILGYINT GKQEGAKLLC GGGIAADRGY FIQPTVFGDV QDGMTIAKEE IFGPVMQILK FKTIEEVVGR ANNSTYGLAA AVFTKDLDKA NYLSQALQAG TVWVNCYDVF GAQSPFGGYK MSGSGRELGE YGLQAYTEVK TVTVKVPQKN S

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct detection of usual and atypical alleles on the human aldehyde dehydrogenase-2 (ALDH2) locus
Hsu LC
American Journal of Human Genetics, 41, 996-1001 (1987)
Cloning of cDNAs for human aldehyde dehydrogenases 1 and 2.
Hsu LC
Proceedings of the National Academy of Sciences of the USA, 82, 3771-3775 (1985)
Significant association between decreased ALDH2 activity and increased sensitivity to genotoxic effects in workers occupationally exposed to styrene.
Weng Z, et al.
Oncotarget, 7(25), 38224-38224 (2016)
Evaluation of pleiotropic effects among common genetic loci identified for cardio-metabolic traits in a Korean population.
Kim Y K, et al.
Cardiovascular Diabetology, 15(1), 20-20 (2016)
Overexpression of aldehyde dehydrogenase-2 (ALDH2) transgene prevents acetaldehyde-induced cell injury in human umbilical vein endothelial cells: role of ERK and p38 mitogen-activated protein kinase.
Li SY
The Journal of Biological Chemistry, 279, 11244-11252 (2004)

Questions

1–2 of 2 Questions  
  1. Is this enzyme in active form?

    1 answer
    1. The product is 100 µg of active enzyme expressed in E. coli. The specific activity is lot-dependent.

      Helpful?

  2. does this enzyme come with a cofactor or coenzyme?

    1 answer
    1. This product is provided as the Aldehyde Dehydrogenase Enzyme without a cofactor.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service