Skip to Content
Merck
All Photos(1)

Key Documents

SRP6007

Sigma-Aldrich

NT-3 from mouse

recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)

Synonym(s):

HDNF, NGF-2, NT-3, Nerve growth factor-2, Neurotrophic factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

mouse

recombinant

expressed in E. coli

Assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

mol wt

13.6 kDa

packaging

pkg of 10 μg

impurities

Endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... NTF3(18205)

General description

NT-3 (neurotrophin-3) is a member of the family of trophic factors, called neurotrophins. This family also includes NGF (nerve growth factor), BDNF (brain-derived neurotrophic factor), and NT-4.[1] NT-3 is the most abundantly expressed neurotrophin during early development.[2]
Murine NT-3 produced from Escherichia coli contains 119 amino acid and having a molecular mass is 13.6kDa.

Application

NT-3 (neurotrophin-3) has been used for the ex vivo treatment of skin punches obtained from rat and human samples.[3]

Biochem/physiol Actions

Study in mice show that NT-3 (neurotrophin-3), along with BDNF (brain-derived neurotrohpic factor), is essential for inner ear development.[1] In vitro motor, enteric, primary sensory, sympathetic, locus coeruleus, cerebellar and hippocampal neurons are responsive to NT-3. Mice null for NT-3 show significant deficiency in sympathetic and sensory nerve populations.[2] In mice, post birth up-regulation of NT-3 induces a second wave of dopaminergic amacrine cell genesis.[4]

Physical form

Sterile filtered and lyophilized powder

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in H?O to a concentration of 0.1-1.0 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Severe sensory and sympathetic deficits in mice lacking neurotrophin-3.
Fari?as I et al
Nature, 369(6482), 658-661 (1994)
Complementary roles of BDNF and NT-3 in vestibular and auditory development.
Ernfors P et al
Neuron, 14(6), 1153-1164 (1995)
Development of a pharmacodynamic biomarker to measure target engagement from inhibition of the NGF-TrkA pathway.
Price EA et al
Journal of Neuroscience Methods, 282, 34-42 (2017)
Overexpression of neurotrophin-3 stimulates a second wave of dopaminergic amacrine cell genesis after birth in the mouse retina.
Yoshida M et al
The Journal of Neuroscience, 31(35), 12663-12673 (2011)
Shyan-Yuan Kao et al.
Bone, 145, 115837-115837 (2021-01-02)
The bone encasing the inner ear, known as the otic capsule, is unique because it remodels little postnatally compared to other bones in the body. Previous studies established that osteoprotegerin (OPG) in the inner ear inhibits otic capsule remodeling. OPG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service