Skip to Content
Merck
All Photos(2)

Key Documents

SRP5277

Sigma-Aldrich

LYN A, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FLJ26625, JTK8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

434-657 nmol/min·mg

mol wt

~81 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LYN(4067)

General description

LYN A is a member of the Src-family of protein kinases that participate in signaling pathways of a variety of cell surface receptors and are localized to the cytoplasmic side of the plasma membrane. LYN A is biosynthetically transported to the plasma membrane via the Golgi pool of caveolin along the secretory pathway. The trafficking of LYN A from the Golgi apparatus to the plasma membrane is inhibited by deletion of the kinase domain but not by kinase inactivation. The kinase domain of LYN A plays a role in LYN A trafficking besides catalysis of substrate phosphorylation. cDNA microarray analysis revealed that LYN A may be involved in the changes that are related to tumor pathogenesis.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service