Skip to Content
Merck
All Photos(2)

Key Documents

SRP5236

Sigma-Aldrich

NDR2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

KIAA0965, STK38L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1.7-2.3 nmol/min·mg

mol wt

~82 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... STK38L(23012)

General description

NDR2 or Nuclear Dbf2-related protein is a serine/threonine kinase in the subgroup of the AGC kinase family. NDR2 is regulated by Mob proteins and is also known as serine/threonine kinase 38 like (STK38L). NDR2 act as novel pro-apoptotic kinase that is activated by RASSF1A/MST1 in response to Fas receptor stimulation and promotes apoptosis. NDR2 is mainly involved in the regulation of structural processes in differentiating and mature neuronal cells and it is highly expressed with highest levels observed in the thymus.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Devroe et al.
The Journal of biological chemistry, 279(23), 24444-24451 (2004-04-07)
Human NDR1 (nuclear Dbf2-related) is a widely expressed nuclear serine-threonine kinase that has been implicated in cell proliferation and/or tumor progression. Here we present molecular characterization of the human NDR2 serine-threonine kinase, which shares approximately 87% sequence identity with NDR1.
T Millward et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(11), 5022-5026 (1995-05-23)
Human, Drosophila melanogaster, and Caenorhabditis elegans cDNA clones encoding homologues of a serine(threonine) protein kinase (EC 2.7.1.37) (designated Ndr protein kinase) have been isolated and sequenced. The human and Drosophila cDNAs predict polypeptides of 54 kDa and 52 kDa, respectively
Anton Vichalkovski et al.
Current biology : CB, 18(23), 1889-1895 (2008-12-09)
Human NDR1 and 2 (NDR1/2) are serine-threonine protein kinases in a subgroup of the AGC kinase family. The mechanisms of physiological NDR1/2 activation and their function remain largely unknown. Here we report that Fas and TNF-alpha receptor stimulation activates human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service