Skip to Content
Merck
All Photos(1)

Key Documents

SRP5166

Sigma-Aldrich

BAX, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~49 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BAX(581)

Related Categories

General description

BCL2 associated X, apoptosis regulator (BAX) is a proapoptotic protein of the B-cell lymphoma 2 (BCL2) protein family. The gene encoding it is localized on human chromosome 19q13.33.[1][2][3]

Biochem/physiol Actions

BCL2 associated X, apoptosis regulator (BAX) forms a heterodimer with B-cell lymphoma 2 (BCL2) and functions as an apoptotic activator. BAX interacts with and increases the opening of the mitochondrial voltage-dependent anion channel (VDAC), which leads to the loss in the mitochondrial membrane potential and the release of cytochrome c. The expression of BAX gene is regulated by the tumor suppressor p53 and BAX has been shown to be involved in p53-mediated apoptosis. Multiple alternatively spliced transcript variants, which encode different isoforms, have been reported for this gene.[2][4][5]

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bcl-2 family proteins and cancer
KW Yip
Oncogene (2008)
Wnt signal transduction pathway and apoptosis: a review
Nives P
Cancer Cell International (2010)
The role of Bcl-2 and its pro-survival relatives in tumourigenesis and cancer therapy
P. N. Kelly
Cell Death and Differentiation (2011)
Role of Bcl-2 family proteins in apoptosis:
apoptosomes or mitochondria?
Yoshihide Tsujimoto
Genes Cells (2001)
S Shimizu et al.
Nature, 399(6735), 483-487 (1999-06-12)
During transduction of an apoptotic (death) signal into the cell, there is an alteration in the permeability of the membranes of the cell's mitochondria, which causes the translocation of the apoptogenic protein cytochrome c into the cytoplasm, which in turn

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service