Skip to Content
Merck
All Photos(1)

Key Documents

SRP5161

Sigma-Aldrich

4EBP1, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

BP-1, EIF4EBP1, MGC4316, PHAS-I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~40 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EIF4EBP1(1978)

General description

Eukaryotic translation initiation factor 4E binding protein 1 (E1F4EBP1) is part of the translation repressor protein family. The gene encoding it is localized on human chromosome 8p11-p12.[1][2]

Biochem/physiol Actions

Eukaryotic translation initiation factor 4E binding protein 1 (E1F4EBP1) is involved in protein synthesis, cell growth and survival. In the unphosphorylated form, it suppresses the translation initiation of capped mRNAs, whereas in phosphorylated form, it increases translation initiation. Phosphorylation of E1F4EBP1 releases it from eukaryotic translation initiation factor 4E (eIF4E) and this results in cap-dependent translation. The protein is also involved in tumorigenesis and the gene is overexpressed in various carcinomas. It is also a substrate for mTORC1 (mammalian target of rapamycin complex 1). In non-phosphorylated conditions, E1F4EBP1 interacts with p21 (cyclin-dependent kinase inhibitor) and enhances destabilization of p21.[3][1][1]

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MicroRNAs 125a and 125b inhibit ovarian cancer cells through post-transcriptional inactivation of EIF4EBP1.
Lee M
Oncotarget (2016)
Stabilization of p21 by mTORC1/4E-BP1 predicts clinical outcome of head and neck cancers.
Llanos S
Nature Communications (2016)
Multiple interacting oncogenes on the 8p11-p12 amplicon in human breast cancer.
Yang ZQ
Cancer Research (2006)
A Pause et al.
Nature, 371(6500), 762-767 (1994-10-27)
The cloning is described of two related human complementary DNAs encoding polypeptides that interact specifically with the translation initiation factor eIF-4E, which binds to the messenger RNA 5'-cap structure. Interaction of these proteins with eIF-4E inhibits translation but treatment of
K Tsukiyama-Kohara et al.
Genomics, 38(3), 353-363 (1996-12-15)
Two related eukaryotic initiation factor-4E binding proteins (4E-BP1 and 4E-BP2) were recently characterized for their capacity to bind specifically to eIF4E and inhibit its function. Here, we determined the cDNA sequence, tissue distribution, genomic structure, and chromosome localization of murine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service