Skip to Content
Merck
All Photos(2)

Documents

SRP5093

Sigma-Aldrich

TLK2 (388-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC44450, PKU-α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥80% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

67-91 nmol/min·mg

mol wt

~67 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TLK2(11011)

General description

TLK2 or Tousled-like kinase 2, is a nuclear serine/threonine kinases that is thought to be involved in the regulation of chromatin assembly. In addition, TLK2 is also involved in other cellular processes such as intracellular signaling cascade, protein amino acid phosphorylation, cell cycle, chromatin modification and in response to DNA damage stimulus. TLK2 displays maximal activity during S phase of the cell cycle where it appears to be regulated by cell-cycle-dependent phosphorylation. Inhibition of DNA replication causes a rapid loss of TLK2 activity, indicating that TLK2 function is tightly linked to ongoing DNA replication.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Yamakawa et al.
Gene, 202(1-2), 193-201 (1998-01-14)
We have cloned cDNAs for novel serine/threonine protein kinases (PK), termed PKU-alpha and PKU-beta, by screening a bacteriophage expression library for kinase activity. Sequence analysis of PKU-alpha and PKU-beta genes revealed that their open reading frames (ORF) were 2151 and
H H Silljé et al.
The EMBO journal, 18(20), 5691-5702 (1999-10-16)
The Tousled (TSL) gene of the plant Arabidopsis thaliana encodes a serine/threonine kinase that is essential for proper flower development. Here we report the cloning and characterization of two human putative homologues of the Arabidopsis TSL gene, termed TLK1 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service