Skip to Content
Merck
All Photos(2)

Key Documents

SRP5084

Sigma-Aldrich

ROS1 (1883-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MCF3, ROS, c-ros-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

544-736 nmol/min·mg

mol wt

~82 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ROS1(6098)

General description

ROS1 is a proto-oncogene and member of the sevenless subfamily of tyrosine kinase insulin receptor genes. ROS1 is highly-expressed in a variety of tumor cell lines and functions as a growth or differentiation factor receptor. The FIG gene can fuse with the ROS1 gene in glioblastoma cell lines. The resulting ROS1/FIG fusion protein is a constitutively activated tyrosine kinase. Direct interaction of ROS1 and the phosphatase SHP-1 can lead to efficient downregulation of ROS1-mediated signaling. Binding sites in the ROS1 cytoplasmic domain display high affinity binding to the SHP-1 N-terminal SH2 domain.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alain Charest et al.
Genes, chromosomes & cancer, 37(1), 58-71 (2003-03-28)
The transmembrane proto-oncogene receptor tyrosine kinase (RTK) ROS is an orphan receptor that is aberrantly expressed in neoplasms of the central nervous system. Here, we report the fusion of its carboxy-terminal kinase domain to the amino-terminal portion of a protein
Christoph Biskup et al.
Journal of cell science, 117(Pt 21), 5165-5178 (2004-10-01)
Signaling of receptor tyrosine kinases (RTKs) is regulated by protein-tyrosine phosphatases (PTPs). We previously discovered the efficient downregulation of Ros RTK signaling by the SH2 domain PTP SHP-1, which involves a direct interaction of both molecules. Here, we studied the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service