Skip to Content
Merck
All Photos(2)

Documents

SRP5082

Sigma-Aldrich

QIK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp434K1115, KIAA0781, LOH11CR1I, SIK2, SNF1LK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

95-129 nmol/min·mg

mol wt

~150 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SIK2(23235)

General description

QIK is a serine/threonine protein kinase that contains an N-terminal kinase domain, a central domain with ubiquitin-associate motif, and a C-terminal PKA phosphorylation site. QIK can phosphorylate IRS1 and overexpression of QIK in adipocyte elevates the phosphorylation of IRS1. The QIK-mediated phosphorylation of IRS1 may modulate the efficiency of insulin signal transduction and could be responsible for insulin resistance associated with diabetes. Insulin disrupts TORC2 activity by induction of QIK which then stimulates the phosphorylation and cytoplasmic translocation of TORC2. Phosphorylated TORC2 is subsequently degraded by the 26S proteasome.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renaud Dentin et al.
Nature, 449(7160), 366-369 (2007-09-07)
During feeding, increases in circulating pancreatic insulin inhibit hepatic glucose output through the activation of the Ser/Thr kinase AKT and subsequent phosphorylation of the forkhead transcription factor FOXO1 (refs 1-3). Under fasting conditions, FOXO1 increases gluconeogenic gene expression in concert
Nanao Horike et al.
The Journal of biological chemistry, 278(20), 18440-18447 (2003-03-08)
Salt-inducible kinase (SIK), first cloned from the adrenal glands of rats fed a high salt diet, is a serine/threonine protein kinase belonging to an AMP-activated protein kinase family. Induced in Y1 cells at an early stage of ACTH stimulation, it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service