Skip to Content
Merck
All Photos(2)

Key Documents

SRP5030

Sigma-Aldrich

FGFR2 (285-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

BFR-1, CD332, CEK3, ECT1, JWS, K-SAM, TK14, TK25

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥80% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

100-136 nmol/min·mg

mol wt

~72 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FGFR2(2263)

General description

FGFR2 is a member of the fibroblast growth factor receptor family which play a role in mitogenesis and differentiation. FGFR2 is a high-affinity receptor for acidic, basic and/or keratinocyte growth factor, and mutations in FGFR2 are associated with Crouzon syndrome, Pfeiffer syndrome, Craniosynostosis, Apert syndrome, Jackson-Weiss syndrome, Saethre-Chotzen syndrome, and syndromic craniosynostosis. FGFR2 is required for early postimplantation development between implantation and the formation of the egg cylinder. FGFR2 contributes to the outgrowth, differentiation, and maintenance of the inner cell mass.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-hsin Kan et al.
American journal of human genetics, 70(2), 472-486 (2002-01-10)
It has been known for several years that heterozygous mutations of three members of the fibroblast growth-factor-receptor family of signal-transduction molecules-namely, FGFR1, FGFR2, and FGFR3-contribute significantly to disorders of bone patterning and growth. FGFR3 mutations, which predominantly cause short-limbed bone
E Arman et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(9), 5082-5087 (1998-06-06)
We disrupted the fibroblast growth factor (FGF) receptor 2 (FGFR2) gene by introducing a neo cassette into the IIIc ligand binding exon and by deleting a genomic DNA fragment encoding its transmembrane domain and part of its kinase I domain.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service