Skip to Content
Merck
All Photos(1)

Key Documents

SRP4893

Sigma-Aldrich

TFF1 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

BCEI, HP1, HPS, TFF-1, TFF1, pS2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~8.2 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TFF1(7031)

General description

Trefoil factor 1(TFF1) is a member of trefoil factor family (TFF) and is encoded by the gene mapped to human chromosome 21q22.3.[1] Recombinant human TFF1 is an 8.2kDa polypeptide containing 60 amino acid residues. TFF1 consists of trefoil motif, which is a 40-amino acid domain with three conserved disulfides.[2] TFF1 is expressed and secreted by epithelial cells in the stomach or colon mucous membrane.[3]

Biochem/physiol Actions

Trefoil factor family (TFF) members play a vital role in mucus stabilization and stimulation of normal epithelial cell restitution during wound healing via motogenic and antiapoptotic activities.[2] TFF1 prevents anoikis, chemicals or bad-induced apoptosis by decreasing the caspase-3, caspase-6, caspase-8, and caspase-9 activities.[1] TFF1 functions as a tumor suppressor in gastric cancer, whereas it acts as a tumor promoter in mucinous ovarian cancer via regulating the Wnt/β-catenin signaling pathway.[2] Elevated level of TFF1 is observed in patients suffering from Chronic kidney disease (CKD). TFF1 might function as a biomarker for early detection of CKD.[4]

Physical form

Sterile filtered and lyophilized powder.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. The solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trefoil factor 1 elevates the malignant phenotype of mucinous ovarian cancer cell through Wnt/?-catenin signaling.
Zhao S
International Journal of Clinical and Experimental Pathology, 8(9), 10412-10419 (2015)
Elena Romano et al.
International journal of molecular sciences, 19(7) (2018-07-13)
Trefoil factor 1 (TFF1) is a small secreted protein expressed in the gastrointestinal tract where, together with the other two members of its family, it plays an essential role in mucosal protection and repair against injury. The molecular mechanisms involved
Saichit Khummuang et al.
Biological procedures online, 19, 14-14 (2017-11-21)
Human trefoil factor (TFF) peptides consist of three members: TFF1, TFF2 and TFF3. TFF3 is the most abundant TFF peptide in saliva. TFF3 homodimer was suggested to be involved in apoptosis inhibition and malignancy. Determination of TFF3 homodimer expression profiles
Functional expression of recombinant human trefoil factor 1 by Escherichia coli and Brevibacillus choshinensis.
Cheng YM
BMC biotechnology, 15:32 (2015)
Trefoil Factor 1 Excretion Is Increased in Early Stages of Chronic Kidney Disease.
Lebherz-Eichinger D
PLoS ONE, 10(9), e0138312-e0138312 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service