Skip to Content
Merck
All Photos(1)

Key Documents

SRP4838

Sigma-Aldrich

PTEN human

recombinant, expressed in E. coli

Synonym(s):

5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN, Mutated in multiple advanced cancers 1, PTEN, Phosphatase and tensin homolog, Phosphatidylinositol-3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90%

form

liquid

mol wt

~86.0 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.1 EU/μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTEN(5728)

Biochem/physiol Actions

PTEN, a tumor suppressor, has been implicated in a large number of human tumors and is conserved from humans to worms. Characterization of PTEN protein showed that it is a phosphatase that acts on proteins and on 3-phosphorylated phosphoinositides, and can therefore modulate signal-transduction pathways that involve lipid second messengers. Recent results indicate that at least part of its role is to regulate the activity of the serine/threonine kinase AKT/PKB, and thus influence cell survival signaling. Recombinant PTEN is purified by proprietary chromatographic techniques.

Physical form

Sterile filtered liquid at 100 μg/ml.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tasneem Bawa-Khalfe et al.
Oncotarget, 8(11), 17651-17664 (2016-11-17)
SUMO protease SENP1 is elevated in multiple carcinomas including prostate cancer (PCa). SENP1 exhibits carcinogenic properties; it promotes androgen receptor-dependent and -independent cell proliferation, stabilizes HIF1α, increases VEGF, and supports angiogenesis. However, mice expressing an androgen-responsive promoter driven SENP1-transgene (SENP1-Tg)
Bjoern C Froehlich et al.
Proteomics. Clinical applications, 14(5), e2000034-e2000034 (2020-07-10)
Immuno-MALDI (iMALDI) combines immuno-enrichment of biomarkers with MALDI-MS for fast, precise, and specific quantitation, making it a valuable tool for developing clinical assays. iMALDI assays are optimized for the PI3-kinase signaling pathway members phosphatase and tensin homolog (PTEN) and PI3-kinase
Sarah A Altınoğlu et al.
Biomaterials science, 4(12), 1773-1780 (2016-10-18)
The phosphatase and tensin homolog deleted on chromosome 10 (PTEN) is a tumor suppressor, mutated or inactive in a large percentage of human cancers. Restoring PTEN activity in cancer cells through gene therapy has shown to inhibit cell growth and
Mohammad Imran Khan et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(3), 3198-3211 (2018-11-01)
Growth factor receptor-binding protein 10 (GRB10) is a well-known adaptor protein and a recently identified substrate of the mammalian target of rapamycin (mTOR). Depletion of GRB10 increases insulin sensitivity and overexpression suppresses PI3K/Akt signaling. Because the major reason for the
Junchao Cai et al.
Nature cell biology, 20(4), 465-478 (2018-03-30)
The contribution of autophagy to cancer development remains controversial, largely owing to the fact that autophagy can be tumour suppressive or oncogenic in different biological contexts. Here, we show that in non-small-cell lung cancer (NSCLC), casein kinase 1 alpha 1 (CK1α) suppresses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service