Skip to Content
Merck
All Photos(1)

Key Documents

SRP4121

Sigma-Aldrich

IGF-I from rat

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

IGF-I, IGF1, IGFIA, Somatomedin C

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~7.69 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Igf1(24482)

General description

IGF-I (insulin-like growth factor-I) is a member of a family of polypeptide growth factors. Rat IGF-I is a 7.69kDa protein containing 70 amino acid residues.
IGF-I exists in two isoforms (IGF1A and IGF1B) in mouse due to alternative transcription.[1]

Application

IGF-I (Insulin-like Growth Factor-I) from rat has been used to study the the effect of IGF-I, IL-27 (interleukin-27) and IL-35 on the blood flow in the inflamed joints of rats.[2]
IGF-I from rat has also been used to test its effect on the expression of its receptor in rheumatoid arthritis model.[3]

Biochem/physiol Actions

IGF-I (Insulin-like growth factor-I) is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types including muscle, bone and cartilage tissue. It mainly mediates growth and development. IGF-I is required for fetal growth and development.[4] It is also involved in the growth and repair of heart tissues.[5] IGF-I has been linked to neuroplasticity and hippocampal neurogenesis.[6] It is important in the recovery from ischaemic injury.[7]
IGF-I interacts with interleukins IL-27 and IL-35 and may play a role in chronic joint inflammation.[3]

Physical form

Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Glucose metabolism and hepatic Igf1 DNA methylation are altered in the offspring of dams fed a low-salt diet during pregnancy.
Siqueira FR
Physiology & Behavior, 154, 68-75 (2016)
The effect of interleukins 27 and 35 and their role on mediating the action of insulin Like Growth Factor -1 on the inflammation and blood flow of chronically inflamed rat knee joint.
Houshmandi N
Cytokine, 81, 117-126 (2016)
Stefano Schiaffino et al.
Skeletal muscle, 1(1), 4-4 (2011-07-30)
A highly conserved signaling pathway involving insulin-like growth factor 1 (IGF1), and a cascade of intracellular components that mediate its effects, plays a major role in the regulation of skeletal muscle growth. A central component in this cascade is the
Elham Abdi et al.
Iranian journal of immunology : IJI, 15(1), 14-27 (2018-03-20)
IGF-1 and certain other cytokines have been shown to exert inflammatory/anti-inflammatory roles in chronic joint diseases. To assess the effect of IGF-1, IL-27 and IL-35, their interaction and their receptor expression in a rheumatoid arthritis model. Freund's adjuvant-induced chronic joint
María Llorens-Martín et al.
The Neuroscientist : a review journal bringing neurobiology, neurology and psychiatry, 15(2), 134-148 (2009-03-25)
This review addresses the role of serum insulin-like growth factor 1 (IGF1) as one mechanism of adult neural plasticity, specifically, its regulation of hippocampal neurogenesis among other plasticity-related processes. It is suggested that IGF has been reused advantageously both for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service