Skip to Content
Merck
All Photos(1)

Key Documents

SRP4066

Sigma-Aldrich

FGF-21 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

FGF21, FGFL, Fibroblast growth factor 21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

~19.5 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF21(26291)

General description

FGF21 (fibroblast growth factor) belongs to FGF superfamily. It is generated by the liver. The gene is mapped to human chromosome 19q13.33.
Recombinant Human FGF-21 produced in Escherichia coli is a single, non-glycosylated polypeptide chain containing 182 amino acids and having a molecular mass of 19.5kDa.

Biochem/physiol Actions

FGF21 (fibroblast growth factor) is a hormonal factor which controls glucose homeostasis and energy metabolism. It exhibits anti-diabetic properties by controlling glucose uptake in peripheral tissues. It also possesses autocrine effects in white adipose tissue. FGF21 is present in milk and is needed for intestinal function in the neonate. It is also involved in bone loss. FGF21 exhibits hepatoprotective properties.

Physical form

Lyopohilized from 10 mM Tris, pH 7.5 plus 100 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute the lyophilized product with sterile water at a concentration of 0.1 mg/mL, which can be further diluted into other aqueous solutions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel locus including FGF21 is associated with dietary macronutrient intake.
Chu AY
Human Molecular Genetics, 22, 1895-1902 (2013)
A Liver-Bone Endocrine Relay by IGFBP1 Promotes Osteoclastogenesis and Mediates FGF21-Induced Bone Resorption.
Wang X
Cell Metabolism, 22, 811-824 (2015)
Human FGF-21 Is a Substrate of Fibroblast Activation Protein.
Coppage AL
PLoS ONE, 11, e0151269-e0151269 (2016)
Fibroblast Activation Protein Cleaves and Inactivates Fibroblast Growth Factor 21.
Dunshee DR
The Journal of Biological Chemistry, 291, 5986-5996 (2016)
Wen-Pin Huang et al.
Journal of cellular and molecular medicine, 26(8), 2451-2461 (2022-03-22)
Circulating endothelial progenitor cells (EPCs), which function in vascular repair, are the markers of endothelial dysfunction and vascular health. Fibroblast growth factor 21 (FGF21), a liver-secreted protein, plays a crucial role in glucose homeostasis and lipid metabolism. FGF21 has been reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service