Skip to Content
Merck
All Photos(1)

Key Documents

SRP4053

Sigma-Aldrich

FGF-8 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

AIGF, FGF-8b, FGF8, Fibroblast Growth Factor-8, HBGF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized

potency

≥2x10^6 units/mg

mol wt

~22.4 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF8(2253)

General description

Fibroblast growth factor-8 (FGF-8) is also known as androgen-induced growth factor (AIGF). It is a heparin-binding growth factor. The gene encoding this protein is localized on human chromosome 10.[1] Recombinant human FGF-8 is a 22.4kDa protein containing 193 amino acid residues.

Biochem/physiol Actions

Fibroblast growth factor-8 (FGF-8) stimulates the proliferation and activation of cells that express the FGF receptors. It has a role in embryogenesis. FGF8b, an isoform of FGF-8, is expressed in prostate cancer.[2][1]

Physical form

Lyophilized in 10 mM Tris, pH 8.0 plus 75 mM NaCl. The protein may appear as a haze or film, which may not be seen at the bottom of the vial.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in 5 mM Tris, pH 8.0 to a concentration of 0.1-1 mg/mL. This solution can be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karolina Jendryczko et al.
Frontiers in pharmacology, 12, 748936-748936 (2021-12-07)
Targeted therapies are a promising alternative to conventional chemotherapy, with an increasing number of therapeutics targeting specific molecular aberrancies in cancer cells. One of the emerging targets for directed cancer treatments is fibroblast growth factor receptors (FGFRs), which are known
A novel locus of ectodermal dysplasia maps to chromosome 10q24.32-q25.1.
Rafiq MA
The Journal of Investigative Dermatology, 124(2), 338-342 (2005)
Screening a phage display library for a novel FGF8b-binding peptide with anti-tumor effect on prostate cancer.
Wang W
Experimental Cell Research, 319(8), 1156-1164 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service