Skip to Content
Merck
All Photos(1)

Key Documents

SRP3317

Sigma-Aldrich

ANGPTL-3 human

recombinant, expressed in CHO cells, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

ANG-5, ANGPT5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in CHO cells

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

62.0 kDa

packaging

pkg of 50 μg

impurities

endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ANGPTL3(27329)

General description

ANGPTL-3 (Angiopoietin like protein 3) is a member of the angiopoietin family of structurally related proteins, characterized by a coiled N-terminal domain and a C-terminal fibrinogen like domain.[1] It is mainly present in the hepatocytes and is also secreted into circulation.[2] The gene is mapped to human chromosome 1p31.[3]
Recombinant human ANGPTL-3 is a glycoprotein that migrates by SDS-PAGE (sodium dodecyl sulfate polyacrylamide gel electrophoresis) analysis at an apparent molecular weight of 62kDa, and contains 452 amino acid residues including a C-terminal His tag.

Biochem/physiol Actions

ANGPTL-3 (Angiopoietin like protein 3) can exert activities related to both angiogenesis and lipid metabolism.[4][5] ANGPTL-3 inhibits lipoprotein lipase (LPL) and endothelial lipase (EL), which has the effect of increasing plasma levels of triglycerides and HDL (high-density lipoprotein)-associated cholesterol.[2] The fibrinogen like portion of the ANGPTL-3 protein can bind α-5/β-3 integrins leading to endothelial cell adhesion and migration.[5] Mutations in this gene can result in familial combined hypobetalipoproteinemia.[1]

Physical form

Lyophilized from 10mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Zhao et al.
Cardiovascular diabetology, 15(1), 132-132 (2016-09-14)
Angiopoietin-like protein 3 (ANGPTL3) is a major lipoprotein regulator and shows positive correlation with high-density lipoprotein-cholesterol (HDL-c) in population studies and ANGPTL3 mutated subjects. However, no study has looked its correlation with HDL components nor with HDL function in patients
Circulating angiopoietin-like protein 8 (ANGPTL8) and ANGPTL3 concentrations in relation to anthropometric and metabolic profiles in Korean children: a prospective cohort study.
Chung HS, et al.
Cardiovascular Diabetology, 15, 1-1 (2016)
ANGPTL3 stimulates endothelial cell adhesion and migration via integrin alpha vbeta 3 and induces blood vessel formation in vivo.
Camenisch G, et al.
The Journal of Biological Chemistry, 277, 17281-17281 (2002)
Post-GWAS methodologies for localisation of functional non-coding variants: ANGPTL3.
Oldoni F, et al.
Atherosclerosis, 246, 193-193 (2016)
Mitsuru Ono et al.
The Journal of biological chemistry, 278(43), 41804-41809 (2003-08-12)
Angiopoietin-like 3 (ANGPTL3) is a secreted protein that is mainly expressed in the liver and regulates lipid metabolism by inhibiting the lipolysis of triglyceriderich lipoproteins. Using deletion mutants of human ANGPTL3, we demonstrated that the N-terminal coiled-coil domain-containing fragment-(17-207) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service