Skip to Content
Merck
All Photos(1)

Key Documents

SRP3310

Sigma-Aldrich

IL-1ALPHA human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

Endogenous Pyrogen (EP), Hematopoietin-1, Lymphocyte-activating factor (LAF)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.001 ng/mL

mol wt

18.0 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL1A(3552)

General description

IL-1α (interleukin) is one of the three isoforms of IL-1, the other two being IL-1β and IL-1Rα. It is a proinflammatory macrophage cytokine. IL-1α is produced de novo and can either be actively secreted or can be released passively by apoptotic cells. This gene is localized to human chromosome 2q13–2q14.1.
Recombinant human IL-1α is an 18.0 kDa protein containing 159 amino acid residues.

Biochem/physiol Actions

IL-1α (interleukin) and IL-1β bind with IL-1 receptor and function as proinflammatory cytokines, whereas IL-1Rα competes to bind with IL-1R but is not proinflammatory. IL-1α can function as an intracrine molecule or as a transcription factor. IL-1 plays a major role in ischemic brain damage, as the inactivation of both IL-1α and IL-1β lead to significant suppression in brain injury. However, inactivation of only IL-1α does not have much influence on overall damage due to some compensatory modifications in the IL-1 system. A hospital-based case-control study showed that rs3783553 variant in this gene is linked with reduced risk of breast cancer. Tumor cell production of IL-1α helps their transmigration across the endothelium, and the expression of IL-1α is linked with distant metastasis in patients with head and neck squamous cell carcinoma.

Physical form

Lyophilized from 3.2 mM Tris, pH 8.3 + 16 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of IL-1alpha and IL-1beta in ischemic brain damage.
Boutin H et al
The Journal of Neuroscience, 21(15), 5528-5534 (2001)
Macrophage cytokines: involvement in immunity and infectious diseases.
Arango Duque G and Descoteaux
Frontiers in Immunology, 5(491) (2014)
Starnes, T. et al.
Journal of Immunology, 2, 642- 646 (2002)
A functional insertion/deletion polymorphism in the IL1A gene is associated with decreased risk of breast cancer.
Huang X et al
Genetics and molecular research : GMR, 15(1), gmr-gmr (2016)
Associations of the interleukin-1 gene locus polymorphisms with risk to hip and knee osteoarthritis: gender and subpopulation differences.
Kaarvatn MH at al
Scandinavian Journal of Immunology, 77(2), 151-161 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service