Skip to Content
Merck
All Photos(1)

Key Documents

SRP3285

Sigma-Aldrich

GREMLIN-1 human

recombinant, expressed in CHO cells, ≥90% (SDS-PAGE), ≥90% (HPLC)

Synonym(s):

CKTSF1B1, DAND2, DRM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in CHO cells

Assay

≥90% (HPLC)
≥90% (SDS-PAGE)

form

lyophilized

potency

0.07-0.11 μg/mL

mol wt

13.7 kDa

packaging

pkg of 50 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GREM1(26585)

General description

Gremlin-1 (isoform-1) belongs to a group of diffusible proteins which bind to ligands of the transforming growth factor-β (TGF-β) family and regulate their activity by inhibiting their access to signaling receptors. The gene encoding this protein is localized on human chromosome 15q13.3.[1] Recombinant Gremlin-1 is a 13.7kDa protein containing 160 amino acid residues.

Biochem/physiol Actions

The interplay between transforming growth factor-β (TGF-β) ligands and their natural antagonists has major biological significance during development processes, in which cellular response can vary considerably depending upon the local concentration of the signaling molecule. Gremlin is highly expressed in the small intestine, fetal brain, and colon and lower expression in brain, prostate, pancreas and skeletal muscle. Gremlin-1 regulates multiple functions in early development by specifically binding to and inhibiting the function of bone morphogenetic proteins -2,-4,-7 (BMP-2, -4, and -7). The protein has a role in limb and digit formation and in nephrogenesis. It also plays a role in carcinogenesis and kidney branching morphogenesis.[1][2][3]

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5 + 150 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gremlin1 and chronic pancreatitis: a new clinical target and biomarker?
Brazil DP
Journal of Molecular Medicine, 93(10), 1057-1060 (2015)
Common genetic variants at the CRAC1 (HMPS) locus on chromosome 15q13.3 influence colorectal cancer risk.
Jaeger E
Nature Genetics, 40(1), 26-28 (2008)
Meta-analysis Reveals Genome-Wide Significance at 15q13 for Nonsyndromic Clefting of Both the Lip and the Palate, and Functional Analyses Implicate GREM1 As a Plausible Causative Gene.
Ludwig KU
PLoS Genetics, 12(3), e1005914-e1005914 (2016)
W Shi et al.
American journal of physiology. Lung cellular and molecular physiology, 280(5), L1030-L1039 (2001-04-06)
Bone morphogenetic protein-4 (BMP-4) is a key morphogen for embryonic lung development that is expressed at high levels in the peripheral epithelium, but the mechanisms that modulate BMP-4 function in early mouse lung branching morphogenesis are unclear. Here, we studied
Dong Qin et al.
Experimental and therapeutic medicine, 19(3), 2353-2359 (2020-02-28)
Proliferative vitreoretinopathy (PVR) is characterised by the contraction and growth of fibrotic membranes on the retina and within the vitreous body. Retinal pigment epithelial (RPE) cells, a major cellular component of the fibrotic membrane, is one of the cell types

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service