Skip to Content
Merck
All Photos(1)

Key Documents

SRP3256

Sigma-Aldrich

TPO from rat

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

MGDF, Megakaryocyte colony-stimulating factor, Thrombopoietin, c-MPL Ligand

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.2 ng/mL ED50

mol wt

18.7 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... THPO(81811)

General description

TPO (thrombopoietin) is a hematopoietic cytokine expressed in multiple tissue types. It is predominantly produced by liver and kidney and acts as a ligand for the cell surface receptor c-mpl.[1][2]

Biochem/physiol Actions

TPO (thrombopoietin) is the key regulator of thrombopoiesis for platelet lineage. In both mice and humans, inactivation of this gene results in significant decrease in number of mature platelets produced by marrow progenitors. Transgenic mice with increased expression of this protein show progressive fibrosis and osteosclerosis of the bone marrow, as well as histological changes in kidneys, leading to mesangioproliferative glomerulopathy.[1][2]

Sequence

SPVPPACDPR LLNKLLRDSY LLHSRLSQCP DVNPLSIPVL LPAVDFSLGE WKTQTEQSKA QDILGAVSLL LEGVMAARGQ LEPSCLSSLL GQLSGQVRLL LGALQGLLGT QLPPQGRTTA HKDPSALFLS LQQLLRGKVR FLLLVEGPAL CVRRTLPTTA VPSRTSQLLT LNKF

Physical form

Lyophilized from Sodium Citrate, pH 4.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcriptional regulation of bone marrow thrombopoietin by platelet proteins.
McIntosh B and Kaushansky K
Experimental Hematology (2008)
Chronic thrombopoietin overexpression induces mesangioproliferative glomerulopathy in mice.
Shimoda HK
American Journal of Hematology (2007)
Kuter, D.J., et al.
Cytokine Reference, 965-982 (2001)
Krasimira Rozenova et al.
Blood, 125(11), 1730-1738 (2015-02-01)
Hematopoietic stem cell (HSC) self-renewal and multilineage reconstitution are controlled by positive and negative signaling cues with perturbations leading to disease. Lnk is an essential signaling adaptor protein that dampens signaling by the cytokine thrombopoietin (Tpo) to limit HSC expansion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service