Skip to Content
Merck
All Photos(1)

Key Documents

SRP3100

Sigma-Aldrich

KGF human

Animal-component free, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

Fibroblast Growth Factor-7, HBGF-7, Keratinocyte Growth Factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

≤10 ng/mL ED50

mol wt

18.9 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

off-white to yellow

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF7(2252)

General description

Fibroblast growth factor-7 (FGF-7) is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities. The gene encoding this protein is localized on human chromosome 15q21.1.[1][2]

Biochem/physiol Actions

Fibroblast growth factor (FGF) family members are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. FGF7 is a potent epithelial cell-specific growth factor, whose mitogenic activity is predominantly exhibited in keratinocytes but not in fibroblasts and endothelial cells. This protein associates with and activates fibroblast growth factor receptors (FGFRs). Studies of mouse and rat homologs of this gene implicated roles in morphogenesis of epithelium, re-epithelialization of wounds, hair development and early lung organogenesis.[3][4]

Sequence

MCNDMTPEQM ATNVNCSSPE RHTRSYDYME GGDIRVRRLF CRTQWYLRID KRGKVKGTQE MKNNYNIMEI RTVAVGIVAI KGVESEFYLA MNKEGKLYAK KECNEDCNFK ELILENHYNT YASAKWTHNG GEMFVALNQK GIPVRGKKTK KEQKTAHFLP MAIT

Physical form

Lyophilized from 15 mM Sodium Phosphate pH 7.0 + 0.25 M NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in 1x PBS to a concentration of 0.5-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Jing Zhu et al.
PLoS genetics, 10(10), e1004687-e1004687 (2014-10-21)
Epidermal stratification of the mammalian skin requires proliferative basal progenitors to generate intermediate cells that separate from the basal layer and are replaced by post-mitotic cells. Although Wnt signaling has been implicated in this developmental process, the mechanism underlying Wnt-mediated
The Fibroblast Growth Factor signaling pathway.
Ornitz DM and Itoh N
Wiley Interdisciplinary Reviews. Systems Biology and Medicine, 4(3), 215-266 (2015)
A family-based association study after genome-wide linkage analysis identified two genetic loci for renal function in a Mongolian population.
Park H
Kidney International, 83(2), 285-292 (2013)
Laxmi Yeruva et al.
mBio, 5(3), e01241-e01214 (2014-06-26)
It is not currently possible to predict the probability of whether a woman with a chlamydial genital infection will develop pelvic inflammatory disease (PID). To determine if specific biomarkers may be associated with distinct chlamydial pathotypes, we utilized two Chlamydia
FGF7/KGF regulates autophagy in keratinocytes: A novel dual role in the induction of both assembly and turnover of autophagosomes.
Belleudi F
Autophagy, 10(5), 803-821 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service