Skip to Content
Merck
All Photos(1)

Documents

SRP3034

Sigma-Aldrich

E-Selectin human

recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

CD62E, Endothelial leukocyte adhesion molecule 1 (ELAM1),, SELE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in CHO cells

sterility

sterile

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

65-85 kDa

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SELE(6401)

General description

Selectins are a family of calcium-dependent type 1 transmembrane proteins. Endothelial (E)-selectin is a heavily glycosylated transmembrane protein expressed by activated endothelial cells in microvascular linings. E-selectin consists of an N-terminal type 1 lectin domain, an EGF-like domain, 6 sushi (CCP/SCR) domains, a transmembrane sequence, and a short cytoplasmic domain. Recombinant human E-selectin is a 58.6kDa protein containing 535 amino acid residues, corresponding to the extracellular portion of the full length protein. Due to glycosylation, E-selectin migrates at an apparent molecular weight of approximately 65-85kDa by SDS-PAGE analysis under reducing conditions. The gene encoding this protein is localized on human chromosome1q24.2.

Biochem/physiol Actions

E-selectin, along with P-selectin and L-selectin, initiate recruitment of circulating leukocytes from blood to sites of inflammation in the vascular lining through interaction with specific cell surface associated carbohydrate determinants. It is a pro-angiogenic factor. This protein has also been implicated in the pathogenesis of stroke and atherosclerosis.

Sequence

WSYNTSTEAM TYDEASAYCQ QRYTHLVAIQ NKEEIEYLNS ILSYSPSYYW IGIRKVNNVW VWVGTQKPLT EEAKNWAPGE PNNRQKDEDC VEIYIKREKD VGMWNDERCS KKKLALCYTA ACTNTSCSGH GECVETINNY TCKCDPGFSG LKCEQIVNCT ALESPEHGSL VCSHPLGNFS YNSSCSISCD RGYLPSSMET MQCMSSGEWS APIPACNVVE CDAVTNPANG FVECFQNPGS FPWNTTCTFD CEEGFELMGA QSLQCTSSGN WDNEKPTCKA VTCRAVRQPQ NGSVRCSHSP AGEFTFKSSC NFTCEEGFML QGPAQVECTT QGQWTQQIPV CEAFQCTALS NPERGYMNCL PSASGSFRYG SSCEFSCEQG FVLKGSKRLQ CGPTGEWDNE KPTCEAVRCD AVHQPPKGLV RCAHSPIGEF TYKSSCAFSC EEGFELHGST QLECTSQGQW TEEVPSCQVV KCSSLAVPGK INMSCSGEPV FGTVCKFACP EGWTLNGSAA RTCGATGHWS GLLPTCEAPT ESNIP

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven R Barthel et al.
Expert opinion on therapeutic targets, 11(11), 1473-1491 (2007-11-22)
Inflammation and cancer metastasis are associated with extravasation of leukocytes or tumor cells from blood into tissue. Such movement is believed to follow a coordinated and sequential molecular cascade initiated, in part, by the three members of the selectin family
Granger DN
Inflammation and the Microcirculation (2010)
Wiese, G., et al.
Journal of Visualized Experiments, 11, 1009-1009 (2009)
Remodeling of the lectin-EGF-like domain interface in P- and L-selectin increases adhesiveness and shear resistance under hydrodynamic force.
Phan UT
Nature Immunology, 7(8), 883-889 (2006)
Two new susceptibility loci 1q24.2 and 11p11.2 confer risk to severe acne.
He L
Nature Communications, 5, 2870-2870 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service