Skip to Content
Merck
All Photos(1)

Key Documents

SRP2148

Sigma-Aldrich

EF1α, GST tagged human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, FLJ25721, LENG7, PTI1, eEF1A-1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$465.00

$465.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
$465.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

$465.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

frozen liquid

mol wt

~77.3 kDa

packaging

pkg of 10 μg

concentration

350 μg/mL

color

colorless to clear

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EEF1A1(1915)

Biochem/physiol Actions

The alpha subunit of elongation factor-1 (EEF1A) is involved in the binding of aminoacyl-tRNAs to 80S ribosomes. During the process, GTP is hydrolyzed into GDP. To perform this function, EEF1A has domains that bind guanine nucleotides, 80S ribosomes, and aminoacyl-tRNAs. Also, EEF1A interacts with the beta subunit of EEF1 to exchange bound GDP for GTP. Multimolecular complexes involving the eukaryotic elongation factor 1A (eEF1A) have been suggested to play an important role in the channeling (vectorial transfer) of tRNA during protein synthesis HIV-1 matrix protein (MA) was found to interact with elongation factor 1-α (EF1α), an essential component of the translation machinery that delivers aminoacyl-tRNA to ribosomes. The interaction between MA and EF1α impairs translation in vitro, a result consistent with a previously proposed model in which inhibition of translation by the accumulation of Gag serves to release viral RNA from polysomes, permitting the RNA to be packaged into nascent virions. It has been identified that the translation elongation factor, eEF1A, and its guanine nucleotide exchange factor, eEF1Bβ, as translationally controlled tumor protein (TCTP)-interacting partners.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cans, C., et al.
Proceedings of the National Academy of Sciences of the USA, 24, 13892-13897 (2003)
J H Brands et al.
European journal of biochemistry, 155(1), 167-171 (1986-02-17)
The primary structure of the alpha subunit of elongation factor 1 (EF-1 alpha) from human MOLT 4 cells was determined by cDNA sequencing. The data show that the conservation of the amino acid sequence is more than 80% when compared
A Cimarelli et al.
Journal of virology, 73(7), 5388-5401 (1999-06-11)
Human immunodeficiency virus type 1 (HIV-1) gag-encoded proteins play key functions at almost all stages of the viral life cycle. Since these functions may require association with cellular factors, the HIV-1 matrix protein (MA) was used as bait in a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service