Skip to Content
Merck
All Photos(1)

Key Documents

SRP2082

Sigma-Aldrich

WT-1 (+KTS) human

recombinant, expressed in insect cells, ≥60% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$575.00

$575.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
5 μG
$575.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

$575.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in insect cells

Assay

≥60% (SDS-PAGE)

form

frozen liquid

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

150 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... WT1(7490)

Biochem/physiol Actions

WT-1, the product of Wilms′ tumor suppressor gene Wt1, is a nuclear protein with structural motifs characteristic of transcription factors, including four C-terminal zinc fingers. While different pre-mRNA processing could result in 16 isoforms of the protein, the inclusion or exclusion of exon 5 and three amino acids (KTS) between zinc fingers 3 and 4 largely affects the activity of the WT1 protein. Such a complex post-transcriptional regulation, particularly in splicing, may represent a major regulatory mechanism for tumorigenesis of the Wilms′ tumor. With the inclusion of exon 5, WT1 (KTS+) binds to both DNA and RNA and is RNase but not DNase sensitive. This form also co-localizes with splicing factors in a speckled nuclear particle, suggesting that the WT1 protein may function as both a transcription factor and a splicing regulator.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Kreidberg et al.
Cell, 74(4), 679-691 (1993-08-27)
In humans, germline mutations of the WT-1 tumor suppressor gene are associated with both Wilms' tumors and urogenital malformations. To develop a model system for the molecular analysis of urogenital development, we introduced a mutation into the murine WT-1 tumor
D A Haber et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(21), 9618-9622 (1991-11-01)
The chromosome 11p13 Wilms tumor susceptibility gene WT1 appears to play a crucial role in regulating the proliferation and differentiation of nephroblasts and gonadal tissue. The WT1 gene consists of 10 exons, encoding a complex pattern of mRNA species: four
S H Larsson et al.
Cell, 81(3), 391-401 (1995-05-05)
WT1 is a tumor suppressor gene with a key role in urogenital development and the pathogenesis of Wilms' tumor. Two alternative splice sites in the WT1 transcript allow the gene to encode four proteins. These carry four Krüppel-type zinc fingers

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service