Skip to Content
Merck
All Photos(1)

Key Documents

SRP2071

Sigma-Aldrich

Androgen receptor (650-920), human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

AIS, DHTR, HUMARA, HYSP1, KD, NR3C4, SBMA, SMAX1, TFM

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$646.00

$646.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
$646.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$646.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~33.7 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

300 μg/mL

color

clear
colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AR(367)

Biochem/physiol Actions

The androgen receptor is an androgen-activated member of the nuclear receptor superfamily of transcription factors. Signaling by the androgen receptor plays a key role in proper development and function of male reproductive organs. The androgen receptor gene is more than 90 kb long and codes for a protein that has 3 major functional domains: the N-terminal domain, DNA-binding domain, and androgen-binding domain. The protein functions as a steroid-hormone activated transcription factor. Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. This gene contains 2 polymorphic trinucleotide repeat segments that encode polyglutamine and polyglycine tracts in the N-terminal transactivation domain of its protein. Expansion of the polyglutamine tract causes spinal bulbar muscular atrophy (Kennedy disease). Mutations in this gene are also associated with complete androgen insensitivity (CAIS). Two alternatively spliced variants encoding distinct isoforms have been described. The androgen receptor (AR) has been shown to play a critical role in the development and progression of prostate cancer.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shalini Murthy et al.
The Prostate, 64(4), 362-372 (2005-03-09)
1,25(OH)2D3 inhibits the growth of prostate cancer cells; previous reports suggest that 1,25(OH)2D3 actions in LNCaP prostate cancer cells are androgen dependent. This is due in part to the observation that the androgen receptor (AR) antagonist, Casodex, modestly inhibits LNCaP
Rute B Marques et al.
International journal of cancer, 117(2), 221-229 (2005-05-19)
To study the mechanisms whereby androgen-dependent tumors relapse in patients undergoing androgen blockade, we developed a novel progression model for prostate cancer. The PC346C cell line, established from a transurethral resection of a primary tumor, expresses wild-type (wt) androgen receptor
Eric Metzger et al.
Nature, 437(7057), 436-439 (2005-08-05)
Gene regulation in eukaryotes requires the coordinate interaction of chromatin-modulating proteins with specific transcription factors such as the androgen receptor. Gene activation and repression is specifically regulated by histone methylation status at distinct lysine residues. Here we show that lysine-specific

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service